71019BC
Assurance® GDS MPX ID for Top STEC Kit
suitable for PCR testing in Food and Beverages applications
About This Item
Recommended Products
Product Name
GDS MPX ID for Top STEC, BioControl, Molecular based secondary screening PCR test for the detection and identification of "Top Six" non-O157 Shiga Toxigenic E. coli (STEC) in food and environmental samples
Agency
AOAC® (PTM)
Quality Level
shelf life
6‑12 mo.
packaging
pkg of 52 tests
manufacturer/tradename
Assurance GDS®
BioControl Systems 71019-52
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
for use with GDS Rotor-Gene
storage temp.
2-8°C
suitability
enterovirulent E. coli (EEC) (shiga toxigenic)
General description
Application
Features and Benefits
- Designed specifically to follow GDS MPX Top STEC assays
- Identification of the specific O serogroup(s) responsible for the positive STEC result and verifies that all targets relevant to the definition of a Top 6 STEC (eae, stx and O group) are present within a single organism
- Confirm isolated colonies from samples presumptive positive for Top 6 STEC
- Utilizes the same single-step mEHEC® enrichment media common to all BioControl methods for Salmonella, E.coli O157:H7, Shiga Toxin genes, and Top STEC
- Only 14 hours from start to finish
Components
- MPX Group 1 STEC Amplification Tubes
- MPX Group 2 STEC Amplification Tubes
- Top 6 STEC Concentration Reagent
- Resuspension Buffer Tq
- Top STEC Wash Solution
Compatibility
Testing Method
Legal Information
configured for
required but not provided
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.
Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.
Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.
Assurance® GDS offers integrated solution for fast and accurate food pathogen detection in microbiology testing.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service