Skip to Content
Merck
All Photos(4)

Key Documents

71010-576BC

Millipore

Assurance® GDS Listeria monocytogenes Tq Kit

suitable for PCR testing in Food and Beverages applications

Synonym(s):

Nucleic acid amplification system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106302
NACRES:
NA.85

Product Name

GDS Listeria monocytogenes Tq, BioControl, Molecular-based PCR assay suitable for detection of Listeria monocytogenes in food and environmental samples.

Agency

according to Immunomagnetic Separation (IMS)
meets requirements for AOAC® PTM
meets requirements for Health Canada
meets requirements for MicroVal

Quality Level

product line

Assurance GDS®

shelf life

6-12 mo.

feature

0.1 mL amp tubes

packaging

pkg of 576 tests

technique(s)

RT-PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

configured for BioControl Systems GDS Rotor-Gene
configured for BioControl Systems GDS

storage temp.

2-8°C

suitability

Listeria monocytogenes

General description

Assurance GDS®, genetic detection system, for Listeria monocytogenes Tq is an automated nucleic acid amplification system for the detection of Listeria monocytogeness in food and environmental samples. The proprietary PickPen® IMS-based sample preparation procedure quickly and effectively captures Listeria monocytogenes, separating the pathogen from inhibitory background materials ensuring valid PCR reactions for all sample types, including challenging food and environmental samples. Assurance GDS® PCR technology provides two additional levels of specificity through the use of both highly specific primers and probes to ensure accurate detection of Listeria monocytogenes.

Application

GDS, genetic detection system, for Listeria monocytogenes Tq is an automated nucleic acid amplification system for the detection of Listeria monocytogenes in foods, ingredients, and environmental samples.

Features and Benefits

  • Results in 24 hours for environmental samples and select foods
  • Single-step enrichment reduces handling time
  • Validated for liquid milk, Mexican soft cheese, frankfurters, deli turkey, raw fish, raw green beans, stainless steel, rubber, concrete, plastic, among others
  • Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and faster time to results
  • Simplified sample preparation does not require heating blocks or preparation of reagents

Components

Each 576 test kit includes:
  • Listeria monocytogenes Amplification Tubes Tq (0.1 mL)
  • Listeria Concentration Reagent
  • Listeria Resuspension Buffer Tq
  • Wash Solution


NOTE: The 100 kit includes standard 0.2 mL amp tubes whereas the 576 kit includes smaller 0.1 mL amp tubes.  The 576 kit requires additional equipment tools to process per the kit Directions for Use.

Compatibility

BioControl® GDS

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

AOAC is a registered trademark of AOAC International
Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
BioControl is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service