Skip to Content
Merck
All Photos(1)

Key Documents

70072

Sigma-Aldrich

Ethylenediaminetetraacetic acid tripotassium salt dihydrate

puriss. p.a., ≥99.0% (KT)

Synonym(s):

EDTA tripotassium salt, Edathamil, Tripotassium ethylenediaminetetraacetate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(KOOCCH2)2NCH2CH2N(CH2COOK)CH2COOH · 2H2O
CAS Number:
Molecular Weight:
442.54
Beilstein:
6762497
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

Assay

≥99.0% (KT)

form

solid

mp

182 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤20 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤3000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

functional group

amine
carboxylic acid

SMILES string

[K+].[K+].[K+].[H]O[H].[H]O[H].OC(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.3K.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;2*1H2/q;3*+1;;/p-3

InChI key

MAPFUJCWRWFQIY-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Justin P Dassie et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 22(11), 1910-1922 (2014-06-24)
Cell-targeted therapies (smart drugs), which selectively control cancer cell progression with limited toxicity to normal cells, have been developed to effectively treat some cancers. However, many cancers such as metastatic prostate cancer (PC) have yet to be treated with current

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service