Skip to Content
Merck
All Photos(2)

Key Documents

69669

Supelco

Leucomalachite Green

analytical standard

Synonym(s):

4,4′-Benzylidenebis(N,N-dimethylaniline)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[C6H4N(CH3)2]2
CAS Number:
Molecular Weight:
330.47
Beilstein:
2140506
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

100-102 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CN(C)c1ccc(cc1)C(c2ccccc2)c3ccc(cc3)N(C)C

InChI

1S/C23H26N2/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,23H,1-4H3

InChI key

WZKXBGJNNCGHIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Leucomalachite green is a triphenylmethane dye and a metabolite of malachite green.

Application

Leucomalachite green may be used as an analytical reference standard for the quantification of the analyte in fish samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of malachite green and leucomalachite green in edible goldfish muscle by liquid chromatography?ion trap mass spectrometry
Lee C-K, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 843(2), 247-251 (2006)
Sara L Stead et al.
Analytical chemistry, 82(7), 2652-2660 (2010-03-06)
A robust screening assay employing solid phase extraction (SPE) followed by a novel aptamer-based procedure is presented for the rapid detection and semiquantitation of the triphenylmethane dye, Malachite Green (MG) and its primary metabolite Leucomalachite Green (LMG) in fish tissue.
Juliana C Hashimoto et al.
Journal of AOAC International, 95(3), 913-922 (2012-07-24)
A simple method using LC/MS/MS was developed and validated to determine residues of malachite green (MG) and leucomalachite green (LMG) in fish fillet. A modified QuEChERS (Quick, Easy, Cheap, Effective, Rugged, and Safe) technique was used to perform the sample
Kamila Mitrowska et al.
Journal of chromatography. A, 1207(1-2), 94-100 (2008-09-06)
A liquid chromatography with visible and fluorescence detection (LC-vis/FLD) method for screening and a liquid chromatography with mass spectrometry (LC-MS/MS) method for the confirmation of malachite green (MG) and its major metabolite, leucomalachite green (LMG) residues in water have been
Chaoyang Long et al.
Journal of chromatography. A, 1203(1), 21-26 (2008-07-29)
With a new oxidant for post-column chemical derivation, a novel approach was developed for the determination of Malachite Green (MG) and Leucomalachite Green (LMG) in fish by high-performance liquid chromatography (HPLC). A C(8) column was used for separation, and elution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service