Skip to Content
Merck
All Photos(2)

Key Documents

67268

Supelco

Myricitrin

analytical standard

Synonym(s):

3,3′,4′,5,5′,7-Hexahydroxyflavone 3-O-rhamnoside, Myricetin 3-O-α-L-rhamnopyranoside, Myricetin 3-O-rhamnoside, Myricitroside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4cc(O)c(O)c(O)c4)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C21H20O12/c1-6-14(26)17(29)18(30)21(31-6)33-20-16(28)13-9(23)4-8(22)5-12(13)32-19(20)7-2-10(24)15(27)11(25)3-7/h2-6,14,17-18,21-27,29-30H,1H3/t6-,14-,17+,18+,21-/m0/s1

InChI key

DCYOADKBABEMIQ-OWMUPTOHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Myricitrin is a plant compound, isolated from Folium Rhododendri Micranthi, which acts as a nitric oxide and protein kinase inhibitor and is found to exhibit antipsychotic-like effect in animal models.

Application

Myricitrin may be used as an analytical reference standard for determination of the analyte in Folium Rhododendri Micranthi extract using high-performance liquid chromatography. It may also be used as an external standard in the validation of both commercial herbal medicines and crude material of Myrcia uniflora extracts using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a HPLC method for standardization of herbal and commercial extracts of Myrcia uniflora
Revista Brasileira de Farmacognosia, 21(3), 402-406 (2011)
Flavia Carla Meotti et al.
Free radical biology & medicine, 44(1), 109-120 (2007-10-30)
Flavonoids are increasingly being ingested by the general population as chemotherapeutic and anti-inflammatory agents. They are potentially toxic because of their conversion to free radicals and reactive quinones by peroxidases. Little detailed information is available on how flavonoids interact with
Ching-Huai Ko et al.
Molecular cancer therapeutics, 4(2), 281-290 (2005-02-17)
Colorectal carcinoma is a leading cause of human mortality due to its high metastatic ability. Because the activation of matrix metalloproteinases (MMP) is a key factor in the metastatic process, agents with the ability to inhibit MMP activity have potential
Jeffrey R Jones et al.
Journal of natural products, 74(1), 38-44 (2010-12-15)
Target-based drug discovery for Alzheimer's disease (AD) centered on modulation of the amyloid β peptide has met with limited success. Therefore, recent efforts have focused on targeting the microtubule-associated protein tau. Tau pathologically accumulates in more than 15 neurodegenerative diseases
M Pereira et al.
Progress in neuro-psychopharmacology & biological psychiatry, 35(7), 1636-1644 (2011-06-22)
Myricitrin is a nitric oxide (NO) and protein kinase C (PKC) inhibitor that has central nervous system activity, including anxiolytic-like action. Nitric oxide inhibitors blocked the behavioral effects of apomorphine, suggesting an antipsychotic-like effect. Furthermore, PKC inhibition reduced psychotic symptoms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service