Skip to Content
Merck
All Photos(3)

Key Documents

66930

Sigma-Aldrich

N-Methyl-D-glucamine

ReagentPlus®, ≥99.0% (T)

Synonym(s):

1-Deoxy-1-(methylamino)-D-glucitol, Meglumine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO5
CAS Number:
Molecular Weight:
195.21
Beilstein:
385906
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

biological source

synthetic

Quality Level

product line

ReagentPlus®

Assay

≥99.0% (T)

form

powder

optical activity

[α]20/D −16.5±0.5°, c = 2% in H2O

autoignition temp.

~662 °F

mp

126-131 °C
129-131.5 °C (lit.)

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤20 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CNC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C7H17NO5/c1-8-2-4(10)6(12)7(13)5(11)3-9/h4-13H,2-3H2,1H3/t4-,5+,6+,7+/m0/s1

InChI key

MBBZMMPHUWSWHV-BDVNFPICSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyl-D-glucamine, also known as Meglumine, is a sorbitol-derived amino sugar. It acts as a catalyst in one-pot synthesis of pyranopyrazole derivatives.

Application

N-Methyl-D-glucamine is utilized as:
  • An ion exchange resin due to its high affinity and selectivity for specific ions
  • A buffering agent to remove boron from an aqueous solution

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymeric microspheres with N-methyl-d-glucamine ligands for boron removal from water solution by adsorption?membrane filtration process.
Wolska J
Environmental Geochemistry and Health, 32(4), 349-352 (2010)
Synthesis of graft adsorbent with N-methyl-D-glucamine for boron adsorption.
Hoshina H
Journal of ion exchange, 18(4), 236-239 (2007)
Separation of aceclofenac and diclofenac in human plasma by free zone capillary electrophoresis using N-methyl-D-glucamine as an effective electrolyte additive
A Zinellu, et al.
European Journal of Pharmaceutical Sciences, 24, 375-380 (2005)
Water-insoluble polymer?clay nanocomposite ion exchange resin based on N-methyl-D-glucamine ligand groups for arsenic removal.
Urbano BF
Reactive functional Polymers, 72(9), 642-649 (2012)
Affinity and selectivity of immobilized N-methyl-D-glucamine for mercury (II) ions.
Zhu X
Industrial & Engineering Chemistry Research, 44(19), 7490-7495 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service