Skip to Content
Merck
All Photos(2)

Key Documents

64947

Supelco

Prochloraz

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16Cl3N3O2
CAS Number:
Molecular Weight:
376.67
Beilstein:
8155344
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCN(CCOc1c(Cl)cc(Cl)cc1Cl)C(=O)n2ccnc2

InChI

1S/C15H16Cl3N3O2/c1-2-4-20(15(22)21-5-3-19-10-21)6-7-23-14-12(17)8-11(16)9-13(14)18/h3,5,8-10H,2,4,6-7H2,1H3

InChI key

TVLSRXXIMLFWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prochloraz is a fungicide possessing imidazole moiety, which can show anti-androgenic activity in blocking androgens like testosterone from mediating their biological effects in the body.[1] It is involved in the inhibition of cytochrome P450-dependent, 14α-demethylase activity required in the conversion of lanosterol to ergosterol, a key constituent of fungal cell membranes.[1]
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antiandrogenic effects in vitro and in vivo of the fungicide prochloraz
Vinggaard MA, et al.
Toxicological Sciences, 69(2), 344-353 (2002)
Karen L Thorpe et al.
Aquatic toxicology (Amsterdam, Netherlands), 105(3-4), 412-420 (2011-08-13)
Exogenous treatment of fish with natural sex hormones and their mimics has been shown to influence gonadal differentiation resulting in biased phenotypic sex-ratios. This has lead to the development of the Fish Sexual Development Test (FSDT) as a method for
Frederik Knud Nielsen et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(2), 343-350 (2011-12-27)
The effects of three model endocrine disruptors, prochloraz, ketoconazole and genistein on steroidogenesis were tested in the adrenocortical H295R cell line to demonstrate that a broader mechanistic understanding can be achieved in one assay by applying chemical analysis to the
Henrik Holbech et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 155(2), 407-415 (2011-11-26)
Results are presented from a validation (with 5 laboratories) of the Fish Sexual Development Test (FSDT) developed to detect endocrine disrupters (EDs) and included in the OECD (Organisation for Economic Co-operation and Development) working program. The aromatase-inhibiting fungicide prochloraz was
Maj-Britt A Bjergager et al.
Aquatic toxicology (Amsterdam, Netherlands), 101(2), 412-422 (2011-01-11)
Laboratory experiments have shown that azole fungicides enhance the toxic effect of pyrethroid insecticides towards the aquatic crustacean Daphnia magna. Due to their sorptive properties the pesticides may, however, be less bioavailable in natural environments, possibly rendering them less toxic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service