Skip to Content
Merck
All Photos(1)

Key Documents

64003BC

Millipore

MVP ICON® Surface Sampling Device

BioControl, ATP Surface Sampling Swab for HACCP Management

Synonym(s):

ATP swab tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112601
NACRES:
NB.79

Quality Level

product line

FLASH®

packaging

pkg of 100 units (swabs; BC64003-100TST)

manufacturer/tradename

MVP ICON®

storage condition

room temperature

application(s)

food and beverages
personnel monitoring (monitoring)
surface monitoring

compatibility

for use with MVP ICON instrument

detection method

bioluminescent

storage temp.

2-30°C
room temp

General description

The MVP ICON® Surface Sampling Device is a self-contained device for use with the MVP ICON® system for quality and HACCP-related monitoring of the environment. The sampling device does not puncture seals during activation, but lets the reagent flow down the swab shaft, and uniformly releases ATP sample from the tip. The MVP ICON® Surface Sampling Device is designed to compensate for variations in swabbing technique, thus offering accurate and consistent readings every time.
The MVP ICON® system detects food and microbiological residues by measuring Adenosine triphosphate (ATP) using luciferase bioluminescence. ATP is found in most food residues and all bacteria, yeast, and mold cells.

Application

MVP ICON® Surface Sampling Device is used with the MVP ICON® system to detect food and microbiological residues during hygiene monitoring for the Food and Beverage industries.

Features and Benefits

Increase Confidence in Your Results
  • Superior swab design eliminates variation in operator′s collection technique, increasing reproducibility and providing assurance swab will be accurately and consistently measured each time.
  • Light output of the reagent is measured, not the solid device surface, ensuring a high degree of repeatability.
  • Reagent flows through hollow sampling device shaft to evenly wash ATP off sampling tip into bottom of the collection tube, ensuring a high degree of accuracy.
  • Sampling devices feature one-hand, one-step activation freeing one hand for other operations.

Create More Flexibility in Your Schedule
  • The swab′s proprietary neutralizing buffer minimizes potential sanitizer interference so measurements can be taken before OR after sanitizing, letting you decide when it is most convenient to conduct your testing.
  • Minimizes potential effect of commonly used sanitizers such as: Quat, Chlorine, and Peracetic Acid.
  • Reduces false negative and false positive results caused by the quenching or amplification of sanitizers on competing swabs.

Convenient Storage
  • Store sampling devices at room temperature – no refrigeration needed; saving precious refrigeration space for other necessities.

Components

All test reagents are contained within the sampling device.

Suitability

ATP Testing, Food Processing, HACCP Monitoring, Sanitation

Principle

The detection of ATP through bioluminescence is a well-established and widely used indicator of surface hygiene and cleaning efficacy in food processing. Visual inspection is not an adequate critical control for surface hygiene and the lengthy time-to-result of microbiological test methods (from 2 - 7 days) make them ineffective for hygiene monitoring. ATP detection with the MVP ICON provides results in 10 seconds allowing for immediate "go/no-go" decisions to be made prior to the start of processing and allows for correction actions, when necessary, to be taken immediately. ATP is a molecule that is found in all living cells, including: bacteria, yeast, mold and all organic matter. After cleaning and/or sanitizing, food processing equipment and the plant environment is tested for ATP. If cleaning/sanitizing has been effective, ATP levels should be low.

Testing Method

ATP Bioluminescence

Storage and Stability

Store at 2 to 30 °C (Protect from direct sunlight. Unused devices must be resealed in the foil bags.)

Legal Information

FLASH is a registered trademark of Merck KGaA, Darmstadt, Germany
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • Swab Tube

  • Wet Chamber

  • Dry Chamber

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service