Skip to Content
Merck
All Photos(1)

Key Documents

63260

Sigma-Aldrich

Sodium maleate dibasic

purum p.a., anhydrous, ≥98.0% (NT)

Synonym(s):

Maleic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH=CHCOONa
CAS Number:
Molecular Weight:
160.04
Beilstein:
6443880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0% (NT)

Quality Level

grade

purum p.a.

Assay

≥98.0% (NT)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Cd: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[Na+].[O-]C(=O)\C=C/C([O-])=O

InChI

1S/C4H4O4.2Na/c5-3(6)1-2-4(7)8;;/h1-2H,(H,5,6)(H,7,8);;/q;2*+1/p-2/b2-1-;;

InChI key

MSJMDZAOKORVFC-UAIGNFCESA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium maleate dibasic is the disodium salt of maleic acid. On reduction in the presence of Raney nickel at 100°C and 2500psi, followed by acidification, it affords succinic acid.1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

C Virgiliou et al.
Bioanalysis, 9(1), 37-51 (2016-12-07)
Till now there is very limited knowledge on the molecular content of coelomic fluid and cells. This study presents the first attempt to elucidate the metabolic profile of such samples. Samples were collected via coelocentesis from 41 women during the
Stephen R Kubota et al.
ChemSusChem, 11(13), 2138-2145 (2018-06-16)
2,5-Furandicarboxylic acid (FDCA) has become an increasingly desirable platform chemical to replace terephthalic acid in the production of a variety of polymeric materials, including polyethylene terephthalate. FDCA can be produced by the oxidation of 5-hydroxymethylfurfural (HMF), which can be derived
Carmen María Ferrer-Luque et al.
Journal of endodontics, 38(6), 826-828 (2012-05-19)
The use of chelating agents in endodontic treatment is required to remove the smear layer and to achieve better disinfection within the root canal system. The aim of this study was to evaluate the residual antimicrobial activity of 17% ethylenediaminetetraacetic
Nidambur Vasudev Ballal et al.
Journal of endodontics, 37(10), 1402-1405 (2011-09-20)
The elimination of microorganisms from the root canal system necessitates the use of combination of irrigating solutions to enhance their antimicrobial property. The combination of irrigants and their interaction sometimes could be detrimental to the outcome of the root canal
Pierangelo Gobbo et al.
Langmuir : the ACS journal of surfaces and colloids, 28(33), 12357-12363 (2012-08-14)
Improved methodology to prepare maleimide-functionalized, water-soluble, small (<3 nm) gold nanoparticles using a retro-Diels-Alder strategy that we developed for similar organic-soluble AuNP's is described. Importantly, our results suggest that a recent paper by Zhu, Waengler, Lennox, and Schirrmacher describing a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service