Skip to Content
Merck
All Photos(2)

Key Documents

63082

Supelco

Magnesium ionophore I

Selectophore, function tested

Synonym(s):

ETH 1117, Magnesium-ligand, N,N′-Diheptyl-N,N′-dimethyl-1,4-butanediamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40N2O2
CAS Number:
Molecular Weight:
340.54
Beilstein:
6408855
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

100
200

product line

Selectophore

form

liquid

quality

function tested

SMILES string

CCCCCCCN(C)C(=O)CCC(=O)N(C)CCCCCCC

InChI

1S/C20H40N2O2/c1-5-7-9-11-13-17-21(3)19(23)15-16-20(24)22(4)18-14-12-10-8-6-2/h5-18H2,1-4H3

InChI key

CBEVANLIGJVSHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium ionophore I (ETH 1117) has been used to prepare calcium-magnesium selective electrode.
Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Neutral synthetic ionophore for Mg2+-selective microelectrodes.; The selectivities as compared to Na+, K+, Ca2+ are sufficient for intracellular measurement of magnesium ion activities: F. Lanter, D. Erne, D. Ammann, W. Simon; Magnesium-selective ionophore for solvent polymeric membrane electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Paper-Based Electronic Tongue.
Witkowska Nery, Emilia, Jessica A. Guimar?es, and Lauro T. Kubota.
Electroanalysis, 27 (10), 2357-2362 (2015)
Xiaolong Xu et al.
Journal of pharmaceutical and biomedical analysis, 165, 338-345 (2018-12-24)
l-ascorbic acid 2-phosphate magnesium (APMg) salt is a vitamin C derivative frequently used in cell culture media for research purposes. It is also used as a raw material in the GMP-manufacturing of gene-, cell- and tissue advanced therapy medicinal products
Erne, D., et al.
Helvetica Chimica Acta, 63, 2271-2271 (1980)
Analytical Chemistry, 52, 2400-2400 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service