Skip to Content
Merck
All Photos(1)

Key Documents

62128

Supelco

(S)-(−)-Limonene

analytical standard

Synonym(s):

(−)-p-Mentha-1,8-diene, (−)-Carvene, (S)-4-Isopropenyl-1-methyl cyclohexene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
2323991
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.7 (vs air)

vapor pressure

<3 mmHg ( 14.4 °C)
1 mmHg ( 20 °C)

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D −94±4°, c = 10% in ethanol

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.1 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.471 (lit.)
n20/D 1.473

bp

175-177 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=C)[C@H]1CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3/t10-/m1/s1

InChI key

XMGQYMWWDOXHJM-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delphine Hardy et al.
Scientific reports, 8(1), 9385-9385 (2018-06-22)
The adult mouse olfactory bulb is continuously supplied with new neurons that mostly differentiate into granule cells (GCs). Different subtypes of adult-born GCs have been identified, but their maturational profiles and their roles in bulbar network functioning and odor behavior
Célia Chenebault et al.
Frontiers in microbiology, 11, 586601-586601 (2020-10-13)
Cyanobacteria, the largest phylum of prokaryotes, perform oxygenic photosynthesis and are regarded as the ancestors of the plant chloroplast and the purveyors of the oxygen and biomass that shaped the biosphere. Nowadays, cyanobacteria are attracting a growing interest in being
Gefu Chi et al.
Inflammation, 36(2), 501-511 (2012-11-28)
The present study aimed to investigate the protective role of limonene in lipopolysaccharide (LPS)-induced acute lung injury (ALI). ALI was induced in mice by intratracheal instillation of LPS (0.5 mg/kg), and limonene (25, 50, and 75 mg/kg) was injected intraperitoneally 1 h prior
Laura Espina et al.
PloS one, 8(2), e56769-e56769 (2013-02-21)
This work explores the bactericidal effect of (+)-limonene, the major constituent of citrus fruits' essential oils, against E. coli. The degree of E. coli BJ4 inactivation achieved by (+)-limonene was influenced by the pH of the treatment medium, being more
Pushpinder Kaur et al.
Natural product communications, 7(9), 1127-1130 (2012-10-19)
A method has been developed for the synthesis of gamma-butyrolactone substituted cinnamyl type Aldol condensation products under milder proline and triethylamine basic conditions. The antioxidant activity of all the synthesized compounds was assessed using three different assays. The cinnamyl type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service