Skip to Content
Merck
All Photos(1)

Key Documents

61767

Sigma-Aldrich

Lectin from Triticum vulgaris

lyophilized, ≥55% (GE), powder

Synonym(s):

WGA, Wheat germ agglutinin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

biological source

Triticum vulgaris

form

powder

quality

lyophilized

concentration

≥55% (GE)

technique(s)

agglutination assay: suitable using fresh human erythrocytes

storage temp.

2-8°C

General description

Lectins are carbohydrate-binding proteins, omnipresent, found in fungi, plants and animals. The structure of lectin is diversely studied in plants and animals. The secondary structure of this protein is rich in β-strands and posses a carbohydrate binding sites on the surface.
WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.

Application

Lectin from Triticum vulgaris has been used in bone resorption assays.
Wheat germ agglutinin conjugated to the outside surface of PLGA nanoparticles loaded with paclitaxel was used to target cancer cells that overexpress WGA-binding glycoproteins (Caco-2 and HT-29 cell lines). WGA-NP showed 50-60% greater endocytosis and intracellular retention of paclitaxel than unlabeled nanoparticles.

Biochem/physiol Actions

Lectin is known to be useful in glycoconjugate characterizing, imaging and targeting. Its use in a microarray assay, enable efficient glycome profiling. This is because of its specific interaction with oligosaccharides, glycoproteins and glycolipids. In plants and fungi, lectin defends against pathogens/feeders. Lectin participates in host recognition and tissue adhesion, thereby aids in the pathogenesis of microorganism.

Other Notes

Molecular characteristics and specificity for sugar binding

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunxia Wang et al.
International journal of pharmaceutics, 400(1-2), 201-210 (2010-09-02)
The purpose of this study was to investigate the potentiation of the anticancer activity and enhanced cellular retention of paclitaxel-loaded PLGA nanoparticles after surface conjugation with wheat germ agglutinin (WGA) against colon cancer cells. Glycosylation patterns of representative colon cancer
Wheat germ agglutinin. Molecular characteristics and specificity for sugar binding.
Y Nagata et al.
The Journal of biological chemistry, 249(10), 3116-3122 (1974-05-25)
Alternative NF-kB Regulates RANKL-Induced Osteoclast Differentiation and Mitochondrial Biogenesis via Independent Mechanisms
Zeng R, et al.
Journal of Bone and Mineral Research, 30(12), 2287-2299 (2015)
Yinglong Zhao et al.
Frontiers in cell and developmental biology, 9, 796902-796902 (2022-02-22)
Microgravity prominently affected cardiovascular health, which was the gravity-dependent physical factor. Deep space exploration had been increasing in frequency, but heart function was susceptible to conspicuous damage and cardiac mass declined in weightlessness. Understanding of the etiology of cardiac atrophy
Rong Zeng et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 30(12), 2287-2299 (2015-06-23)
Mitochondrial biogenesis, the generation of new mitochondrial DNA and proteins, has been linked to osteoclast (OC) differentiation and function. In this study we used mice with mutations in key alternative NF-κB pathway proteins, RelB and NF-κB-inducing kinase (NIK), to dissect

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service