59751
Campylobacter Culture Medium
NutriSelect® Plus, powder, final pH 7.4±0.2 (25 °C), bottle of 500 g, non-sterile, suitable for microbiology
Synonym(s):
Blood Free Campylobacter Broth, Modified CCDA - Preston Broth, Modified Charcoal Cefoperazone Desoxycholate Broth
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
beef extract, 10 g/L
casein enzymic hydrolysate, 3 g/L
charcoal bacteriological, 4 g/L
ferrous sulfate, 0.25 g/L
peptic digest of animal tissue, 1 g/L
sodium chloride, 1 g/L
sodium deoxycholate, 1 g/L
sodium pyruvate, 0.25 g/L
packaging
bottle of 500 g
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
7.4±0.2 (25 °C)
application(s)
environmental
food and beverages
sample preparation
microbiology
suitability
selective for Campylobacter spp.
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.
Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.
Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.
Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service