Skip to Content
Merck
All Photos(1)

Key Documents

59677

Supelco

Methyl dihydrojasmonate, mixture of cis and trans

analytical standard

Synonym(s):

(3-Oxo-2-pentylcyclopentyl)acetic acid methyl ester, Methyl (3-oxo-2-pentylcyclopentyl)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O3
CAS Number:
Molecular Weight:
226.31
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.459 (lit.)

bp

110 °C/0.2 mmHg (lit.)

density

0.998 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCC1C(CCC1=O)CC(=O)OC

InChI

1S/C13H22O3/c1-3-4-5-6-11-10(7-8-12(11)14)9-13(15)16-2/h10-11H,3-9H2,1-2H3

InChI key

KVWWIYGFBYDJQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl dihydrojasmonates (MDJ) are grouped under the class of cyclopentanones. MDJ is a widely used fragrance ingredient in perfume industries because of its intense floral, jasmine-like odor. Both the trans and cis methyl dihydrojasmonate mixture may be isolated from Jasminum grandiflorum L.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective synthesis and absolute stereochemistry of both the enantiomers of trans-magnolione, a fragrance structurally related to trans-methyl jasmonate
Donnoli MI, et al.
Tetrahedron, 60(23), 4975-4981 (2004)
Fragrance material review on methyl dihydrojasmonate
Scognamiglio J, et al.
Food And Chemical Toxicology, 50(23), S562-S571 (2012)
Evaluation of the developmental toxicity of methyl dihydrojasmonate (MDJ) in rats
Politano VT, et al.
International Journal of Toxicology, 27(3), 295-300 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service