Skip to Content
Merck
All Photos(2)

Key Documents

52181

Supelco

Gymnemagenin

analytical standard

Synonym(s):

Olean-12-ene-3β,16β,21β,22α,23,28-hexol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50O6
CAS Number:
Molecular Weight:
506.71
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC1(C)C[C@H]2C3=CC[C@@H]4[C@@]5(C)CC[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]4(C)[C@]3(C)C[C@H](O)[C@@]2(CO)[C@@H](O)[C@@H]1O

InChI

1S/C30H50O6/c1-25(2)13-18-17-7-8-20-26(3)11-10-21(33)27(4,15-31)19(26)9-12-28(20,5)29(17,6)14-22(34)30(18,16-32)24(36)23(25)35/h7,18-24,31-36H,8-16H2,1-6H3/t18-,19+,20+,21-,22-,23-,24-,26-,27-,28+,29+,30-/m0/s1

InChI key

VKJLHZZPVLQJKG-ABHKXHSUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service