Skip to Content
Merck
All Photos(1)

Key Documents

52074

Supelco

Sodium phosphate monobasic

suitable for HPLC, 99.0-101.0% (T)

Synonym(s):

Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98
MDL number:
UNSPSC Code:
41116105
NACRES:
NB.21

Assay

99.0-101.0% (T)

form

powder or crystals

technique(s)

HPLC: suitable

pH

4.0-4.5 (20 °C, 10% in H2O)

solubility

H2O: 0.1 g/mL at 25 °C, clear, colorless

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: ≤0.1
λ: 260 nm Amax: ≤0.06
λ: 280 nm Amax: ≤0.04
λ: 320 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

SMILES string

[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

AJPJDKMHJJGVTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate monobasic may be used as a mobile phase in:
  • High-performance liquid chromatography (HPLC) determination of adenosine in human synovial fluid.
  • Analysis of neuroendocrine tumor markers by HPLC-electrochemical detection.
  • Analysis of benzalkonium chloride and its homologs using HPLC/HPCE.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of benzalkonium chloride and its homologs: HPLC versus HPCE
Prince SJ, et al.
Journal of Pharmaceutical and Biomedical Analysis, 19(6), 877-882 (1999)
HPLC determination of adenosine in human synovial fluid
Sottofattori E, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(5-6), 1143-1146 (2001)
Simultaneous analysis of neuroendocrine tumor markers by HPLC-electrochemical detection
Manickum, T
Journal of Chromatography. B, Biomedical Sciences and Applications, 877(32), 4140-4146 (2009)

Articles

LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.

LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.

LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.

LC-UV-MS workflow details teriparatide peptide mapping, including enzymatic digestion, separation conditions, and QTOF mass spectrometer identification.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service