Skip to Content
Merck
All Photos(1)

Key Documents

51994

Supelco

Cadmium Standard for AAS

TraceCERT®, 1 g/L Cd in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Cd in nitric acid (nominal concentration)

technique(s)

AAS: suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Cadmium standard can be used to prepare standard solution for calibrating the atomic absorption spectrometry instrument and quantifying the levels of cadmium in table salt samples.

Analysis Note

1 g/L Cd in 2% nitric acid, prepared with high purity Cd metal, HNO3 and water

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Muta. 1B - Skin Irrit. 2 - STOT RE 2

Target Organs

Kidney,Bone

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuo Zhang et al.
International journal of molecular sciences, 21(21) (2020-11-04)
Cadmium (Cd) contamination in soils poses great risks to both agricultural production and human health. Calcium (Ca) is an essential element playing a significant role in protecting plants against Cd toxicity. However, how Ca affects Cd uptake and translocation in
Jeffrey D Weidenhamer et al.
Environmental health perspectives, 119(7), 1029-1033 (2011-03-08)
We evaluated the bioavailability of Cd in 86 components of 57 jewelry items found to contain high levels of Cd (> 10,000 ppm) by X-ray fluorescence (XRF), using extractions that simulate mouthing or swallowing of jewelry items. We screened jewelry

Articles

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service