Skip to Content
Merck
All Photos(1)

Key Documents

51393

Sigma-Aldrich

Outlet valve for plastic container 30 L and 60 L

pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

Pricing and availability is not currently available.

packaging

pkg of 1 ea

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
700304725366747408
particle size

5 nm±1 nm (TEM; conforms)

particle size

20 nm (+/- 2nm TEM)

particle size

18-22 nm

particle size

-

density

0.865 g/mL at 25 °C

density

0.865 g/mL at 25 °C

density

1.00 g/mL at 25 °C

density

0.988 g/mL at 25 °C, ~1 g/mL at 25 °C

concentration

5 mg/mL in toluene

concentration

5 mg/mL in toluene

concentration

5 mg/mL in H2O

concentration

1 mg/mL Fe in H2O

magnetization

≥30 emu/g (at ambient temperature under 4500 Oe)

magnetization

>20 emu/g, at room temp. under 4500 Oe

magnetization

>20 emu/g, at 4500Oe

magnetization

>45 emu/g (at room temperature; under 4500 Oe)

avg. part. size

5 nm

avg. part. size

20 nm

avg. part. size

20 nm

avg. part. size

30 nm (TEM)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service