Skip to Content
Merck
All Photos(1)

Key Documents

51363

Supelco

L-Asparagine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
132.12
Beilstein:
1723527
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

235 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

N[C@@H](CC(N)=O)C(O)=O

InChI

1S/C4H8N2O3/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H2,6,7)(H,8,9)/t2-/m0/s1

InChI key

DCXYFEDJOCDNAF-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuo Yanagisawa et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 1), 5-15 (2013-01-01)
Structures of Methanosarcina mazei pyrrolysyl-tRNA synthetase (PylRS) have been determined in a novel crystal form. The triclinic form crystals contained two PylRS dimers (four monomer molecules) in the asymmetric unit, in which the two subunits in one dimer each bind
Christopher J Poon et al.
The Journal of biological chemistry, 280(25), 23615-23621 (2005-04-26)
Keratan sulfate is thought to influence the cleavage of aggrecan by metalloenzymes. We have therefore produced a recombinant substrate, substituted with keratan sulfate, suitable for the study of aggrecanolysis in vitro. Recombinant human G1-G2 was produced in primary bovine keratocytes
Alberto Canepa et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 17(3), 413-421 (2002-02-28)
The aims of this study were to investigate free amino acid (AA) concentrations in plasma, red blood cells (RBC), polymorphonuclear granulocytes (PMN), and muscle, sampled at the same time, in normal and uraemic children. Twelve apparently well-nourished chronically uraemic children
Emma R Job et al.
Journal of immunology (Baltimore, Md. : 1950), 190(5), 2169-2177 (2013-02-01)
Seasonal influenza A viruses (IAV) originate from pandemic IAV and have undergone changes in antigenic structure, including addition of glycans to the viral hemagglutinin (HA). Glycans on the head of HA promote virus survival by shielding antigenic sites, but highly
B Ahlman et al.
Clinical science (London, England : 1979), 86(6), 653-662 (1994-06-01)
1. The effects of short-term starvation and refeeding on the free amino acid concentrations of the intestinal mucosa were characterized in male subjects (n = 6), using endoscopically obtained biopsy specimens from the duodenum and from all four segments of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service