Skip to Content
Merck
All Photos(2)

Key Documents

48280

Sigma-Aldrich

D-(+)-Galacturonic acid monohydrate

≥97% (titration)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O7 · H2O
CAS Number:
Molecular Weight:
212.15
Beilstein:
1727087
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97% (titration)

form

solid

optical activity

[α]20/D +53±2°, 5 hr, c = 10% in H2O

impurities

5-12% water

color

white to off-white

SMILES string

O.OC1O[C@@H]([C@H](O)[C@H](O)[C@H]1O)C(O)=O

InChI

1S/C6H10O7.H2O/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;/h1-4,6-9,12H,(H,10,11);1H2/t1-,2+,3+,4-,6?;/m0./s1

InChI key

BGHPCEJXDOGRGW-KSSASCOMSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitra Arjmandi et al.
Food science and technology international = Ciencia y tecnologia de los alimentos internacional, 23(1), 36-45 (2016-06-28)
The effect of a pasteurization treatment at 90 ± 2 ℃ for 35 s provided by continuous microwave under different doses (low power/long time and high power/short time) or conventional pasteurization on the quality of orange-colored smoothies and their changes throughout 45 days of
Lissa Davis et al.
Journal of food science, 83(7), 1921-1932 (2018-06-16)
Blueberry pomace (BP) and cranberry pomace (CP) are good sources of dietary fiber and phenolics. This study aimed to develop berry fruit pomace (FP)-fortified specialty mustard with elevated bioactive compounds and ascertain consumer acceptance of a new product. Wet BP
Nikita V Penkov
International journal of molecular sciences, 22(21) (2021-11-14)
Despite more than a century of research on the hydration of biomolecules, the hydration of carbohydrates is insufficiently studied. An approach to studying dynamic hydration shells of carbohydrates in aqueous solutions based on terahertz time-domain spectroscopy assay is developed in
Michael A Held et al.
Molecular plant, 4(5), 832-844 (2011-03-23)
Plant cell walls are complex structures that offer structural and mechanical support to plant cells and are ultimately responsible for plant architecture and form. Pectins are a large group of complex polysaccharides of the plant cell wall that are made
Carlo Giannelli et al.
Analytical chemistry, 92(9), 6304-6311 (2020-04-25)
Typhoid fever is a major cause of morbidity and mortality in developing countries. Vaccines based on the Vi capsular polysaccharide are licensed or in development against typhoid fever. Vi content is a critical quality attribute for vaccines release, to monitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service