Skip to Content
Merck
All Photos(3)

Key Documents

46640

Supelco

7-Fluorobenzofurazan-4-sulfonic acid ammonium salt

for HPLC derivatization, LiChropur, ≥98.5% (HPLC)

Synonym(s):

4-Fluoro-7-sulfobenzofurazan ammonium salt, 7-Fluorobenz-2,1,3-oxadiazole-4-sulfonic acid ammonium salt, Ammonium 7-fluorobenzofurazan-4-sulfonate, SBD-F

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3FN2O4S · NH3
CAS Number:
Molecular Weight:
235.19
Beilstein:
5199564
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for HPLC derivatization

Quality Level

Assay

≥98.5% (HPLC)

quality

LiChropur

technique(s)

HPLC: suitable

fluorescence

λex 385 nm; λem 524 nm in 0.1 M borate pH 9.5 (after derivatization with glutathione)

storage temp.

2-8°C

SMILES string

N.OS(=O)(=O)c1ccc(F)c2nonc12

InChI

1S/C6H3FN2O4S.H3N/c7-3-1-2-4(14(10,11)12)6-5(3)8-13-9-6;/h1-2H,(H,10,11,12);1H3

InChI key

JXLHNMVSKXFWAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent probe for thiols and pre-column labeling of biological thiols for HPLC

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Noguchi et al.
Journal of pharmaceutical and biomedical analysis, 10(7), 515-520 (1992-07-01)
A HPLC method is developed for the determination of glutathione isopropyl ester, a drug for the treatment of cerebral vascular disease, in rat, dog and human blood. The blood is deproteinized with sulphosalicylic acid and the clear supernatant treated with
Total plasma homocysteine analysis by HPLC with SBD-F precolumn derivatization.
I Fermo et al.
Methods in molecular biology (Clifton, N.J.), 159, 237-244 (2000-10-29)
I Daskalakis et al.
Biomedical chromatography : BMC, 10(5), 205-212 (1996-09-01)
A sensitive HPLC-fluorescence method for determining total endogenous plasma homocysteine (Hcy), cysteine (Cys) and cysteinylglycine (Cys-Gly) following derivatization with ammonium 7-fluoro 2,1,3-benzoxadiazole-4-sulphonate (SBD-F) is described. Quantitation utilizes an internal standard, 2-mercaptoethylamine. The derivatization procedure has been optimized for concentration of
J B Ubbink
Seminars in thrombosis and hemostasis, 26(3), 233-241 (2000-09-30)
Analytical methods to measure plasma total homocyst(e)ine (tHcy) concentrations are reviewed. High-pressure liquid chromatography (HPLC) with fluorometric detection is the most widely used method to determine plasma tHcy concentrations. Both monobromobimane and ammonium 7-fluorobenzo-2-oxa-1,3-diazole-4-sulphonate (SBD-F) are popular thiol-specific fluorogenic agents
J Tanaka et al.
Journal of biochemistry, 127(4), 597-601 (2000-03-31)
Pseudomonas sp. 109 produces a unique lipase (LipL) which efficiently catalyzes intramolecular transesterification of omega-hydroxyesters to form macrocyclic lactones. The production of the enzymatically active LipL requires a specific molecular chaperon (LimL protein) together with a low-M(r) lipase-activation-factor (LAF) of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service