Skip to Content
Merck
All Photos(1)

Key Documents

46494

Supelco

Nifuroxazide

VETRANAL®, analytical standard

Synonym(s):

5-Nitro-2-furaldehyde p-hydroxybenzoylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N3O5
CAS Number:
Molecular Weight:
275.22
Beilstein:
4264976
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

Oc1ccc(cc1)C(=O)N\N=C\c2ccc(o2)[N+]([O-])=O

InChI

1S/C12H9N3O5/c16-9-3-1-8(2-4-9)12(17)14-13-7-10-5-6-11(20-10)15(18)19/h1-7,16H,(H,14,17)/b13-7+

InChI key

YCWSUKQGVSGXJO-NTUHNPAUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nifuroxazide is an oral antibiotic, which may be used for the treatment of diarrhea.

Application

Nifuroxazide may be used as a reference standard in the determination of the analyte in plasma samples using high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akilimali Shindano et al.
Acta gastro-enterologica Belgica, 70(1), 32-33 (2007-07-11)
We report the case of a middle-aged woman who developed a typical picture of acute pancreatitis together with systemic features of immunoallergy after the intake of two capsules (200 mg) of nifuroxazide. Even if acute pancreatitis is a rare adverse
Mariane B Fernandes et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(3), 535-540 (2012-01-31)
It is important to determine the toxicity of compounds and co-solvents that are used in cell monolayer permeability studies to increase confidence in the results obtained from these in vitro experiments. This study was designed to evaluate the cytotoxicity of
Fávero Reisdorfer Paula et al.
Bioorganic & medicinal chemistry, 17(7), 2673-2679 (2009-03-24)
In this study, in vitro anti-T. cruzi activity assays of nifuroxazide (NX) analogues, such as 5-nitro-2-furfuryliden and 5-nitro-2-theniliden derivatives, were performed. A molecular modeling approach was also carried out to relate the lipophilicity potential (LP) property and biological activity data.
JAK-STAT Pathway in Disease (2009)
Philippe Quillardet et al.
Mutagenesis, 21(5), 305-311 (2006-08-10)
Nitrofurans are widely used in human medicine, as nitrofurantoin and nifuroxazide, still prescribed for long-term antimicrobial prophylaxis of urinary tract and gastrointestinal infection in humans respectively. Recent experiments in mammals, as well as reports mentioning toxic effects in humans associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service