Skip to Content
Merck
All Photos(1)

Key Documents

45400

Supelco

Chlortoluron

PESTANAL®, analytical standard

Synonym(s):

3-(3-Chloro-4-methyl)-1,1-dimethylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClN2O
CAS Number:
Molecular Weight:
212.68
Beilstein:
2647688
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CN(C)C(=O)Nc1ccc(C)c(Cl)c1

InChI

1S/C10H13ClN2O/c1-7-4-5-8(6-9(7)11)12-10(14)13(2)3/h4-6H,1-3H3,(H,12,14)

InChI key

JXCGFZXSOMJFOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Villaverde et al.
Environmental science & technology, 43(21), 8227-8232 (2009-11-21)
This study investigates time-dependent sorption of pesticides in soil aggregates. We tested if the sorption kinetics of pesticides in soil aggregates can be described by modeling diffusion into aggregates for a range of soils and pesticides. Our hypothesis is that
Min-Ling Gao et al.
Journal of environmental sciences (China), 19(3), 327-331 (2007-10-09)
Sorption of chlorotoluron in ammonium sulfate, urea and atrazine multi-solutes system was investigated by batch experiments. The results showed application of nitrogen fertilizers to the soil could affect the behavior of chlorotoluron. At the same concentration of N, sorption of
Sylvie Nélieu et al.
Environmental science & technology, 43(9), 3148-3154 (2009-06-19)
The nitrate-induced photodegradation of chlorotoluron was demonstrated to occur efficiently in natural water through two series of experiments in outdoor aquatic mesocosms. During the first campaign, it was shown that the pesticide degradation kinetics was clearly dependent on nitrate concentration.
Claire Valiente Moro et al.
Environmental toxicology and chemistry, 31(4), 778-786 (2012-01-27)
Extensive use of herbicides in agriculture is accompanied by the risk of environmental contamination of aquatic ecosystems. The present study shows the effects of the herbicides chlortoluron and mesotrione on three microalgae species: two chlorophyceae (Pediastrum tetras, Ankistrodesmus fusiformis) and
Bin Xu et al.
The Science of the total environment, 417-418, 241-247 (2012-01-26)
The degradation of chlortoluron by monochloramination was investigated in the pH range of 4-9. The degradation kinetics can be well described by a second-order kinetic model, first-order in monochloramine (NH(2)Cl) and first-order in chlortoluron. NH(2)Cl was found not to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service