Skip to Content
Merck

44698

Supelco

Bacopaside II

analytical standard

Synonym(s):

Pseudojujubogenin 3-O-[α-L-arabinofuranosyl-(1→2)-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranoside]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H76O18
CAS Number:
Molecular Weight:
929.10
UNSPSC Code:
85151701
NACRES:
NA.24

Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C\C(C)=C/[C@@H]1CO[C@]23C[C@]4(CO2)[C@H](CC[C@@H]5[C@@]6(C)CC[C@H](O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)[C@H]7O[C@@H]9O[C@@H](CO)[C@H](O)[C@H]9O)C(C)(C)[C@@H]6CC[C@@]45C)[C@H]3[C@@]1(C)O

InChI

1S/C47H76O18/c1-21(2)14-22-18-58-47-19-46(20-59-47)23(38(47)45(22,7)57)8-9-28-43(5)12-11-29(42(3,4)27(43)10-13-44(28,46)6)63-41-37(65-39-34(55)31(52)25(16-49)61-39)36(32(53)26(17-50)62-41)64-40-35(56)33(54)30(51)24(15-48)60-40/h14,22-41,48-57H,8-13,15-20H2,1-7H3/t22-,23-,24-,25+,26-,27-,28-,29+,30-,31+,32-,33+,34-,35-,36+,37-,38+,39+,40+,41+,43-,44-,45+,46+,47-/m1/s1

InChI key

WZWPYJOPCULCLQ-CPYMMDDTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bacopaside II is an antidepressant, belonging to the class of triterpenoid saponins isolated from the plant, Bacopa monnieri.[1] Its potential applications include the improvement of high order cognitive processes and treatment/prevention of neurological diseases.[2]

Application

Bacopaside II may be used as an analytical reference standard for the quantification of the analyte in Bacopa monnieri using different chromatography techniques.[2][3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of saponin glycosides in Bacopa monnieri by reversed phase high performance liquid chromatography
Thai Pharmaceutical and Health Science Journal, 2(1), 26-32 (2007)
Separation of bacoside A3 and bacopaside II, major triterpenoid saponins in Bacopa monnieri, by HPTLC and SFC. Application of SFC in implementation of uniform design for herbal drug standardization, with thermodynamic study
Acta Chromatographica , 17(1), 125-125 (2006)
Serotonin and Melatonin: Their Functional Role in Plants, Food, Phytomedicine, and Human Health (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service