44653
EC Broth
suitable for microbiology, NutriSelect® Basic
Synonym(s):
Escherichia Coli Broth
About This Item
Recommended Products
Agency
according to ISO 7251:2005
Quality Level
sterility
non-sterile
form
powder
shelf life
limited shelf life, expiry date on the label
composition
bile salts, 1.5 g/L
casein digest (pancreatic), 20 g/L
dipotassium phosphate, 4 g/L
lactose, 5 g/L
potassium phosphate, 1.5 g/L
sodium chloride, 5 g/L
manufacturer/tradename
NutriSelect® Basic
technique(s)
microbiological culture: suitable
final pH
6.9±0.2 (25 °C)
application(s)
agriculture
bioburden testing
environmental
food and beverages
water monitoring
microbiology
suitability
selective for Escherichia coli
selective for coliforms
Related Categories
Application
Preparation Note
Other Notes
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service