Skip to Content
Merck
All Photos(2)

Key Documents

44426

Supelco

Metanil Yellow

analytical standard

Synonym(s):

3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, Acid Yellow 36

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N3NaO3S
CAS Number:
Molecular Weight:
375.38
Colour Index Number:
13065
Beilstein:
3831568
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[O-]S(=O)(=O)c1cccc(c1)\N=N\c2ccc(Nc3ccccc3)cc2

InChI

1S/C18H15N3O3S.Na/c22-25(23,24)18-8-4-7-17(13-18)21-20-16-11-9-15(10-12-16)19-14-5-2-1-3-6-14;/h1-13,19H,(H,22,23,24);/q;+1/p-1/b21-20+;

InChI key

NYGZLYXAPMMJTE-ANVLNOONSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Metanil Yellow (MY) belongs to the azo group of dyes.

Application

Metanil Yellow (MY) may be used as an analytical reference standard for the determination of MY in:
  • Lipsticks and eye shadows by microwave-assisted dispersive liquid-liquid microextraction (MA-DLLME) using solidification of the floating organic droplet (SFOD) clean-up followed by analysis using high performance liquid chromatography-diode array detection (HPLC-DAD).
  • Ball-point pen ink, in paper ink and in turmeric raw materials as well as finished products by HPLC-DAD.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatography-mass spectrometry and high-performance liquid chromatography-diode array detection for dating of paper ink.
Diaz-Santana O, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 1515, 187-195 (2017)
Microwave-assisted dispersive liquid-liquid microextraction coupling to solidification of floating organic droplet for colorants analysis in selected cosmetics by liquid chromatography.
Zhong Z, et al.
Talanta, 194, 46-54 (2019)
Curcuminoids in Turmeric Roots and Supplements: Method Optimization and Validation.
Mudge E, et al.
Food Analytical Methods, 9(5), 1428-1435 (2017)
Comparison of the main dating methods for six ball-point pen inks.
Diaz-Santana O, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 138, 550-561 (2018)
S Ramchandani et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 32(6), 559-563 (1994-06-01)
The effects of Metanil yellow, Orange II and their blend on hepatic xenobiotic metabolizing enzymes were compared. Parenteral administration of Metanil yellow and Orange II to rats at a dose of 80 mg/kg body weight for 3 days caused a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service