Skip to Content
Merck
All Photos(1)

Key Documents

43530

Supelco

Reserpine Standard for LC-MS

analytical standard, for LC-MS

Synonym(s):

Reserpine, (3β, 16β, 17α, 18β, 20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40N2O9
CAS Number:
Molecular Weight:
608.68
Beilstein:
102014
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:
NACRES:
NB.21

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

5 mg/L in water: isopropyl alcohol (1:1)

technique(s)

LC/MS: suitable

mp

~265 °C (dec.)

suitability

corresponds for mass spectrometry

storage temp.

2-8°C

SMILES string

CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)c6cc(OC)c(OC)c(OC)c6

InChI

1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1

InChI key

QEVHRUUCFGRFIF-MDEJGZGSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reserpine is a 36 carbon indole alkaloid mostly used as a tranquilizing agent in medicine. It is mostly used as drug for hypertension.

Application

It was used as mass recalibration standard in coupling capillary electrophoresis with electrospray ionization time-of-flight mass spectrometry (CE-TOFMS) to overcome the problem of decreasing the sensitivity during the detection of metabolome differential display.

Biochem/physiol Actions

Inhibits vesicular uptake of catecholamines and serotonin.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stimulation of reserpine biosynthesis in the callus of Rauvolfia tetraphylla L. by precursor feeding.
Anitha, S and BD Ranjitha Kumari
African Journal of Biotechnology, 5.8, 659-659 (2006)
Tomoyoshi Soga et al.
The Journal of biological chemistry, 281(24), 16768-16776 (2006-04-13)
Metabolomics is an emerging tool that can be used to gain insights into cellular and physiological responses. Here we present a metabolome differential display method based on capillary electrophoresis time-of-flight mass spectrometry to profile liver metabolites following acetaminophen-induced hepatotoxicity. We
Naomi S Rajapaksa et al.
Organic letters, 15(3), 706-709 (2013-01-22)
A catalytic, enantioselective synthesis of (+)-reserpine is reported. The route features a highly diastereoselective, chiral catalyst-controlled formal aza-Diels-Alder reaction between a 6-methoxytryptamine-derived dihydro-β-carboline and an enantioenriched α-substituted enone to form a key tetracyclic intermediate. This approach addresses the challenge of
Reserpine-induced supersensitivity to the cardiac effects of agonists.
K D Meisheri et al.
Life sciences, 24(6), 473-480 (1979-02-05)
Sandy D Shamon et al.
The Cochrane database of systematic reviews, (4)(4), CD007655-CD007655 (2009-10-13)
Many antihypertensive agents exist today for the treatment of primary hypertension (systolic blood pressure >/=140 mmHg and/or diastolic blood pressure >/=90 mmHg). Randomised controlled trials have been carried out to investigate the evidence for these agents.There is, for example, strong

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service