43348
Farnesol
analytical standard, stabilized, mixture of isomers
Synonym(s):
3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥95.0% (GC)
shelf life
limited shelf life, expiry date on the label
contains
α-tocopherol (synthetic stabilizer)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.488-1.492
n20/D 1.489 (lit.)
bp
149 °C/4 mmHg (lit.)
density
0.886 g/mL at 20 °C (lit.)
application(s)
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care
format
neat
storage temp.
2-8°C
SMILES string
C/C(CC/C=C(C)/CCC=C(C)C)=C\CO
InChI
1S/C15H26O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11,16H,5-6,8,10,12H2,1-4H3/b14-9+,15-11+
InChI key
CRDAMVZIKSXKFV-YFVJMOTDSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
204.8 °F - closed cup
Flash Point(C)
96.00 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service