Skip to Content
Merck
All Photos(3)

Key Documents

42875

Millipore

Hybriscan I kit

Megasphaera, suitable for environmental food and beverages, microbiology and in situ hybriziation

Synonym(s):

Megasphaera HybriScanI, FastScan Megasphaera

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product name

HybriScanI Megasphaera, suitable for environmental, microbiology, suitable for microbe id | in situ hybriziation

technique(s)

microbe id | in situ hybriziation: suitable

Quality Level

application(s)

environmental
microbiology

storage temp.

2-8°C

suitability

Megasphaera spp.

General description

HybriScan I kits are intended for the identification and result confirmation of microorganisms using rRNA as the detection target. Several customers also use it to detect microorganisms in food, beverage and water samples. The principle is based on in situ sandwich hybridization with two specific probes. As the rRNA of dead cells is degraded in the presence of RNase, only living cells are detected. Each cell has several thousand rRNA copies, so neither PCR nor a polymerase is needed for this technology, which means that this molecular biology test is not inhibited by the sample matrix.

Features and Benefits

  • Sensitivity (after pre enrichment): 110 cfu/L
  • Sensitivity (cfu/assay): 5000

Legal Information

HybriScan is a trademark of ScanBec GmbH

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

HybriScan™ assays provide rapid, accurate molecular detection of beer spoilage microorganisms, ensuring excellent product quality in breweries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service