Skip to Content
Merck
All Photos(1)

Key Documents

41144

Supelco

β-Alanine

analytical standard

Synonym(s):

β-Ala, 3-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
906793
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

color

white to off-white

mp

202 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

NCCC(O)=O

InChI

1S/C3H7NO2/c4-2-1-3(5)6/h1-2,4H2,(H,5,6)

InChI key

UCMIRNVEIXFBKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

β-Alanine is a rate-limiting precursor of carnosine, a cytoplasmic dipeptide which is present in human skeletal muscles in high concentrations.

Application

β-Alanine may be used as a reference standard for the determination of β-alanine in:
  • Potato tubers by gas chromatography-mass spectrometry (GC-MS).
  • Human plasma, urine, and cerebrospinal fluid samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS).
  • Striatal microdialysis samples by high performance liquid chromatography-fluorescence detection (HPLC-FLD) with ortho-phthalaldehyde-mercaptoethanol derivatization.
  • Fermented soybean paste, Cheonggukjang, by GC-MS operating in selected-ion monitoring (SIM) mode and principal component analysis (PCA).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Endogenous β-amino acid that is a nonselective agonist at glycine receptors and a ligand for the G protein-coupled orphan receptor, TGR7 (MrgD). β-Alanine is a component of pantothenic acid.
β-Alanine, a β−amino acid, is a component of pantothenic acid and the rate-limiting amino acid in the biosynthesis of the histidinyl antioxidant dipeptides carnosine and anserine. Endogenous β-amino acid that is a nonselective agonist at glycine receptors and a ligand for the G protein-coupled orphan receptor, TGR7 (MrgD). β-Alanine flux plays a cytoprotective role by supporting the osmotic stability of marine organisms, preimplantation mouse embryos and mammalian cells exposed to hypoxic stress.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolite profiling of Cheonggukjang, a fermented soybean paste, during fermentation by gas chromatography-mass spectrometry and principal component analysis
Park MK, et al.
Food Chemistry, 122(4), 1313-1319 (2010)
Simultaneous analysis of metabolites in potato tuber by gas chromatography-mass spectrometry
Roessner U, et al.
The Plant Journal, 23(1), 131-142 (2000)
Rapid and sensitive step gradient assays of glutamate, glycine, taurine and $\gamma$-aminobutyric acid by high-performance liquid chromatography-fluorescence detection with o-phthalaldehyde-mercaptoethanol derivatization with an emphasis on microdialysis samples
Piepponen TP and Skujins A
Journal of Chromatography. B, Biomedical Sciences and Applications, 757(2), 277-283 (2001)
A rapid, sensitive method for quantitative analysis of underivatized amino acids by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
Le A, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 944(1), 166-174 (2014)
?-Alanine supplementation augments muscle carnosine content and attenuates fatigue during repeated isokinetic contraction bouts in trained sprinters
Derave W, et al.
Journal of Applied Physiology, 103(5), 1736-1743 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service