Skip to Content
Merck
All Photos(2)

Key Documents

40922

Supelco

Levofloxacin

analytical standard

Synonym(s):

(−)-Ofloxacin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20FN3O4
CAS Number:
Molecular Weight:
361.37
Beilstein:
7327015
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

optical activity

[α]20/D -104±4° in chloroform

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

OC(C(C1=O)=CN2C(C1=CC(F)=C3N4CCN(C)CC4)=C3OC[C@@H]2C)=O

InChI

1S/C18H20FN3O4/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21/h7-8,10H,3-6,9H2,1-2H3,(H,24,25)/t10-/m0/s1

InChI key

GSDSWSVVBLHKDQ-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abrar K Thabit et al.
International journal of antimicrobial agents, 48(5), 535-541 (2016-11-05)
Increasing antimicrobial resistance in community-acquired pneumonia (CAP) pathogens has contributed to infection-related morbidity and mortality. Delafloxacin is a novel fluoroquinolone with broad-spectrum activity against Gram-positive and -negative organisms, including Streptococcus pneumoniae and methicillin-resistant Staphylococcus aureus (MRSA). This study aimed to
Won-Jung Koh et al.
American journal of respiratory and critical care medicine, 188(7), 858-864 (2013-08-10)
Levofloxacin (LFX) and moxifloxacin (MXF) are the two most frequently recommended fluoroquinolones for treatment of patients with multidrug-resistant tuberculosis (MDR-TB). However, studies comparing the effectiveness of LFX and MXF among patients with MDR-TB are lacking. To compare the effectiveness of
Vanessa R Anderson et al.
Drugs, 68(4), 535-565 (2008-03-06)
Levofloxacin (Levaquin) is a fluoroquinolone antibacterial that is the L-isomer of ofloxacin. A high-dose (750 mg) short-course (5 days) of once-daily levofloxacin is approved for use in the US in the treatment of community-acquired pneumonia (CAP), acute bacterial sinusitis (ABS)
Hans H Liu
Drug safety, 33(5), 353-369 (2010-04-20)
The fluoroquinolone class of antibacterial agents are among the most frequently prescribed drugs, with utility in a broad range of bacterial infections. Although very useful agents, the fluoroquinolones as a class are associated with a number of adverse events, some
John S Bradley et al.
Pediatrics, 134(1), e146-e153 (2014-06-12)
Safety concerns for fluoroquinolones exist from animal studies demonstrating cartilage injury in weight-bearing joints, dependent on dose and duration of therapy. For children treated with levofloxacin or comparator in randomized, prospective, comparative studies for acute otitis media and community-acquired pneumonia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service