Skip to Content
Merck
All Photos(1)

Key Documents

40784

Millipore

Listeria mono Selective Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

Agency

according to ISO 11290-1:2017
according to ISO 11290-2:2017

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

microbiology

storage temp.

2-8°C

Application

A selective supplement recommended by ISO Committee for the isolation of Listeria species.

Components

(per vial, sufficient for 500 ml medium)
Ceftazidime 10.0 mg
Nalidixic acid sodium salt 10.0 mg
Amphotericin B 5.0 mg
Polymyxin B sulfate 38350 IU

Reconstitution

Rehydrate the contents of 1 vial aseptically with 2 ml of 0.2 N Sodium hydroxide, further add 8 ml of sterile distilled water. Mix well and aseptically add it to 465 ml of sterile, molten L. mono Differential Agar Base (Cat. No. 77408) along with sterile contents of Listeria mono Enrichement Supplement I (Cat. No. 03708). Mix well and pour into sterile petri plates.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service