Skip to Content
Merck
All Photos(2)

Key Documents

37094

Supelco

Rapamycin

VETRANAL®, analytical standard

Synonym(s):

Sirolimus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H79NO13
CAS Number:
Molecular Weight:
914.17
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CO[C@@H]1C[C@@H](CC[C@H]1O)C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(C)\[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCC[C@H]4C(=O)O2)OC

InChI

1S/C51H79NO13/c1-30-16-12-11-13-17-31(2)42(61-8)28-38-21-19-36(7)51(60,65-38)48(57)49(58)52-23-15-14-18-39(52)50(59)64-43(33(4)26-37-20-22-40(53)44(27-37)62-9)29-41(54)32(3)25-35(6)46(56)47(63-10)45(55)34(5)24-30/h11-13,16-17,25,30,32-34,36-40,42-44,46-47,53,56,60H,14-15,18-24,26-29H2,1-10H3/b13-11+,16-12+,31-17+,35-25+/t30-,32-,33-,34-,36-,37+,38+,39+,40-,42+,43+,44-,46-,47+,51-/m1/s1

InChI key

QFJCIRLUMZQUOT-HPLJOQBZSA-N

Gene Information

human ... FKBP1A(2280)

General description

Rapamycin belongs to the macrolide class of antibiotics obtained from the bacterium Streptomyces hygroscopicus. It was first used as an anti-fungal agent but later also found use as an immunosuppressant and potential anti-cancer drug. It acts by preventing the activity of serine/threonine kinase and subsequently blocking the cytokine-driven T cell proliferation.

Application

The analytical standard can be used as follows:
  • Electrochemical determination of rapamycin using boron-doped diamond electrode and differential pulse voltammetry (DPV) in human urine, serum, and blood samples
  • Simultaneous determination of six immunosuppressants from whole blood samples by high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) technique combined with modified QuEChERS method

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Complications in Surgery (2006)
B cell activating factor (BAFF) selects IL-10? B cells over IL-10+ B cells during inflammatory responses
Ma N, et al.
Molecular Immunology, 85, 18-26 (2017)
Xing Liu et al.
Journal of virology, 93(17) (2019-06-14)
The Us11 protein encoded by herpes simplex virus 1 (HSV-1) functions to impair autophagy; however, the molecular mechanisms of this inhibition remain to be fully established. Here, we report that the Us11 protein targets tripartite motif protein 23 (TRIM23), which
Carrie L Lucas et al.
Nature immunology, 15(1), 88-97 (2013-10-30)
The p110δ subunit of phosphatidylinositol-3-OH kinase (PI(3)K) is selectively expressed in leukocytes and is critical for lymphocyte biology. Here we report fourteen patients from seven families who were heterozygous for three different germline, gain-of-function mutations in PIK3CD (which encodes p110δ).
A V Tinker et al.
Gynecologic oncology, 130(2), 269-274 (2013-05-16)
HPV infection has been associated with deregulation of the PI3K-Akt-mTOR pathway in invasive cervical carcinomas. This 2-stage phase II study assessed the activity of the mTOR inhibitor, temsirolimus, in patients with measurable metastatic and/or locally advanced, recurrent carcinoma of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service