Skip to Content
Merck
All Photos(1)

Key Documents

36629

Supelco

Atrazine-desethyl

PESTANAL®, analytical standard

Synonym(s):

2-Amino-4-chloro-6-isopropylamino-1,3,5-triazine, Desethylatrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10ClN5
CAS Number:
Molecular Weight:
187.63
Beilstein:
610488
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

100
200

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)Nc1nc(N)nc(Cl)n1

InChI

1S/C6H10ClN5/c1-3(2)9-6-11-4(7)10-5(8)12-6/h3H,1-2H3,(H3,8,9,10,11,12)

InChI key

DFWFIQKMSFGDCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Atrazine-desethyl is found to be a degradation product of the triazine herbicide atrazine.
Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

The Water Dictionary: A Comprehensive Reference of Water Terminology (2011)
Plasma Chemistry and Catalysis in Gases and Liquids (2012)
J Kniewald et al.
Journal of steroid biochemistry, 27(4-6), 1095-1100 (1987-01-01)
Daily s.c. injections of atrazine and deethylatrazine to rat mothers during pregnancy only or during pregnancy and lactation influenced the pituitary-gonadal axis of male and female offsprings. In female and male offspring, slow maturation of gonadotropic system is evident and
E L Arthur et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 32(5), 599-620 (1997-09-01)
The degradation of deethylatrazine (DEA), a major metabolite of atrazine, was studied by using radiotracers in soils with two different atrazine histories. DEA degradation was enhanced in soils which had received long-term exposure to atrazine (atrazine-history soil) compared with soils
T Babić-Gojmerac et al.
Journal of steroid biochemistry, 33(1), 141-146 (1989-07-01)
The inhibitory influence of atrazine and deethylatrazine on testosterone metabolism in male rat anterior pituitary and hypothalamus were studied under in vivo and in vitro experimental conditions. In vivo strong influence of atrazine (12 mg/100 g by wt. daily during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service