Skip to Content
Merck
All Photos(1)

Key Documents

36132

Supelco

Iprodione

PESTANAL®, analytical standard

Synonym(s):

[3-(3,5-Dichlorophenyl)-2,4-dioxoimidazolidinyl]-N-(methylethyl)carboxamide, Iprodione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3N2C3H2O2CONHCH(CH3)2
CAS Number:
Molecular Weight:
330.17
Beilstein:
895003
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

130-134 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CC(C)NC(=O)N1CC(=O)N(C1=O)c2cc(Cl)cc(Cl)c2

InChI

1S/C13H13Cl2N3O3/c1-7(2)16-12(20)17-6-11(19)18(13(17)21)10-4-8(14)3-9(15)5-10/h3-5,7H,6H2,1-2H3,(H,16,20)

InChI key

ONUFESLQCSAYKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iprodione is an agricultural and horticultural dicarboximide fungicide, having potent anti-androgenic property.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feasibility of a microwave liquid sensor based on molecularly imprinted sol-gel polymer for the detection of iprodione fungicide
Bou-Maroun E, et al.
Sensors and Actuators B, Chemical, 244, 24-30 (2017)
Effects of low doses of carbendazim or iprodione either separately or in mixture on the pubertal rat seminiferous epithelium: An ex vivo study
Durand P, et al.
Toxicology in vitro (2017)
Morphogenetic Alterations of Alternaria alternata Exposed to Dicarboximide Fungicide, Iprodione
Kim E, et al.
The plant pathology journal, 33, 95-95 (2017)
Jorge Morales et al.
Chemosphere, 92(11), 1536-1541 (2013-05-21)
The influence of humic aggregates in water solution upon the chemical stability of Iprodione has been investigated under basic conditions. Taking into account that an important part of soils are colloids, the possibility of its presence implies that soil composition
Alexander B Fialkov et al.
Journal of chromatography. A, 1218(52), 9375-9383 (2011-11-29)
A new type of low thermal mass (LTM) fast gas chromatograph (GC) was designed and operated in combination with gas chromatography mass spectrometry (GC-MS) with supersonic molecular beams (SMB), including GC-MS-MS with SMB, thereby providing a novel combination with unique

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service