Skip to Content
Merck
All Photos(1)

Key Documents

36127

Supelco

Climbazole

PESTANAL®, analytical standard

Synonym(s):

1-(4-Chlorophenoxy)-1-(imidazol-1-yl)-3,3-dimethylbutan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17ClN2O2
CAS Number:
Molecular Weight:
292.76
Beilstein:
618020
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

96-100 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C)C(=O)C(Oc1ccc(Cl)cc1)n2ccnc2

InChI

1S/C15H17ClN2O2/c1-15(2,3)13(19)14(18-9-8-17-10-18)20-12-6-4-11(16)5-7-12/h4-10,14H,1-3H3

InChI key

OWEGWHBOCFMBLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Climbazole is an imidazole antifungal agent, which is widely used as an active ingredient in anti-dandruff (AD) shampoos since, it inhibits the microbial growth and improves skin barrier on scalp.

Application

Climbazole may be used as a reference standard in the determination of climbazole in environmental samples and in samples of anti-dandruff shampoo using ultra-high-performance liquid chromatography coupled with tandem mass spectrometry (UHPLC-MS-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of biocides in different environmental matrices by use of ultra-high-performance liquid chromatography--tandem mass spectrometry
Chen F-Z, et al.
Analytical and Bioanalytical Chemistry, 404(10), 3175-3188 (2012)
W Wigger-Alberti et al.
Praxis, 90(33), 1346-1349 (2001-09-06)
Pityrosporum ovale appears to play an important role in the pathogenesis of dandruff as a symptom of seborrheic dermatitis. Climbazole is an antimycotic agent with a high in vitro and in vivo efficacy against P. ovale. In the presented work
A Schmidt
Zentralblatt fur Veterinarmedizin. Reihe B. Journal of veterinary medicine. Series B, 44(4), 193-197 (1997-06-01)
Malassezia pachydermatis is a yeast-like, mainly zoophilic fungus, also known as Malassezia canis. It can be isolated in 20-50% from normal ear cerumen specimen from dogs or cats and has an even higher prevalence in non-suppurative otitis externa in animal
Alex A Pérez-Rivera et al.
Mutation research, 672(1), 27-39 (2008-10-28)
Climbazole is an imidazole antifungal agent that can provide anti-dandruff benefits when incorporated into a shampoo matrix. A series of genotoxicity studies were performed to support the human safety of this azole antifungal drug. Climbazole was not mutagenic in the
C Coiffard et al.
Annales pharmaceutiques francaises, 57(5), 392-396 (1999-10-16)
We compared thermostability of various natural (sulfoprolamine and sodium usnate) or synthetic (climbazol and piroctone olamine) antidandruff agents in aqueous diluted solution at pH around 7. Thermodegradation of these solutions was studied by an isothermal method in thermostatically controlled ovens

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service