Skip to Content
Merck
All Photos(1)

Key Documents

36123

Supelco

Chlorbensid

PESTANAL®, analytical standard

Synonym(s):

4-Chlorobenzyl 4-chlorophenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10Cl2S
CAS Number:
Molecular Weight:
269.19
Beilstein:
2505694
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(CSc2ccc(Cl)cc2)cc1

InChI

1S/C13H10Cl2S/c14-11-3-1-10(2-4-11)9-16-13-7-5-12(15)6-8-13/h1-8H,9H2

InChI key

ZHLKXBJTJHRTTE-UHFFFAOYSA-N

General description

Chlorbensid is an organochlorine pesticide widely used in agriculture.

Application

Chlorbensid may be used as an analytical reference standard for the determination of the analyte in human urine, horticultural samples, water, and soil samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean Boutonnat et al.
BMC cancer, 5, 120-120 (2005-09-21)
PKH67 labelling was compared for classical proliferation assessment (using S phase evaluation) to analyse the cell proliferation of 29 AML patients treated or not with various drugs. Among these drugs, the effect of tetrapeptide AcSDKP or AcSDKP-NH2 on AML cells
Semiautomatic method for the screening and determination of 23 organochlorine pesticides in horticultural samples by gas chromatography with electron-capture detection.
Colume A, et al.
Journal of Chromatography A, 849(1), 235-243 (1999)
A solid phase extraction method for the screening and determination of pyrethroid metabolites and organochlorine pesticides in human urine.
Colume A, et al.
Rapid Communications in Mass Spectrometry, 15(21), 2007-2013 (2001)
On-line solid-phase extraction coupled to supercritical fluid chromatography with diode array detection for the determination of pesticides in water.
Bernal JL, et al.
Journal of Chromatography A, 754(1-2), 145-157 (1996)
Possibilities of gas chromatography-atomic emission detection in pesticide multiresidue analysis Application to herbicide analysis in soils.
Bernal JL, et al.
Journal of Chromatography A, 754(1-2), 245-256 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service