Skip to Content
Merck
All Photos(2)

Key Documents

35495

Supelco

PCB No 180

analytical standard

Synonym(s):

2,2′,3,4,4′,5,5′-Heptachlorobiphenyl, 2,2′,3,4,4′,5,5′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H3Cl7
CAS Number:
Molecular Weight:
395.32
Beilstein:
2509255
Ballschmiter Number:
180
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1cc(Cl)c(cc1Cl)-c2cc(Cl)c(Cl)c(Cl)c2Cl

InChI

1S/C12H3Cl7/c13-6-3-8(15)7(14)1-4(6)5-2-9(16)11(18)12(19)10(5)17/h1-3H

InChI key

WBHQEUPUMONIKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polychlorinated biphenyls have been extensively used as coolants and lubricants in various industries.

Application

PCB No 180 may be used as an analytical reference standard for the determination of the analyte in chicken eggs, human serum and honey samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of pesticides and PCBs in honey by solid-phase extraction cleanup followed by gas chromatography with electron-capture and nitrogen-phosphorus detection.
Herrera A, et al.
Analytical and Bioanalytical Chemistry, 381(3), 695-701 (2005)
Environmental and occupational causes of cancer: a call to act on what we know.
Clapp RW, et al.
Biomedicine and Pharmacotherapy, 61(10), 631-639 (2007)
Rapid multiresidue determination of organochlorine and organophosphorus compounds in human serum by solid-phase extraction and gas chromatography coupled to tandem mass spectrometry.
Pitarch E, et al.
Analytical and Bioanalytical Chemistry, 376(2), 189-197 (2003)
Robert Roos et al.
Toxicology, 284(1-3), 42-53 (2011-04-05)
PCB 180 (2,2',3,4,4',5,5'-heptachlorobiphenyl) is a persistent and accumulating polychlorinated biphenyl abundantly present in food and the environment. In this study, we used highly purified PCB 180 (dioxinlike impurities: 2.7 ng TEQ(WHO)/g PCB 180) in a 28-day toxicity study in young
Roshan Tofighi et al.
Toxicological sciences : an official journal of the Society of Toxicology, 124(1), 192-201 (2011-09-13)
Developmental exposure to food contaminants, such as polychlorinated biphenyls (PCBs), has been considered as a possible cause of neurodevelopmental disorders. We have investigated the effects of noncytotoxic concentrations of PCBs 153 and 180 on spontaneous differentiation of rat embryonic neural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service