Skip to Content
Merck
All Photos(2)

Key Documents

34366

Supelco

Phenoxyacetic acid

PESTANAL®, analytical standard

Synonym(s):

Glycolic acid phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
907949
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

98-100 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC(=O)COc1ccccc1

InChI

1S/C8H8O3/c9-8(10)6-11-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)

InChI key

LCPDWSOZIOUXRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed Ashraf Ali et al.
Bioorganic & medicinal chemistry, 15(5), 1896-1902 (2007-01-26)
A series of 2-{4-[1-amino (thioxo) methyl-5-(substituted phenyl)-4,5-dihydro-1H-3-pyrazolyl]-2-methoxyphenoxy}acetic acid and 2-{4-[1-carbamoyl-5-(substituted phenyl)-4,5-dihydro-1H-3-pyrazolyl]-2-methoxyphenoxy}acetic acid were synthesized and the in vitro activity of the synthesized compounds against Mycobacterium tuberculosis H37Rv (MTB) and INH-resistant M. tuberculosis (INHR-MTB) was studied. Among the synthesized compounds, compound
Karen A Evans et al.
Bioorganic & medicinal chemistry letters, 21(8), 2345-2350 (2011-03-19)
A series of phenoxyacetic acids as subtype selective and potent hPPARδ partial agonists is described. Many analogues were readily accessible via a single solution-phase synthetic route which resulted in the rapid identification of key structure-activity relationships (SAR), and the discovery
N Sundaraganesan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 70(2), 430-438 (2008-02-20)
In this work, we will report a combined experimental and theoretical study on molecular and vibrational structure of o-chlorophenoxy acetic acid (OCPAA) and p-chlorophenoxy acetic acids (PCPAA). The FT-IR and Fourier transform-Raman spectra of both the compounds was recorded in
Tim Luker et al.
Bioorganic & medicinal chemistry letters, 21(12), 3616-3621 (2011-05-20)
A novel series of biaryl phenoxyacetic acids was discovered as potent, selective antagonists of the chemoattractant receptor-homologous expressed on Th2 lymphocytes receptor (CRTh2 or DP2). A hit compound 4 was discovered from high throughput screening. Modulation of multiple aryl substituents
M Shahar Yar et al.
Journal of enzyme inhibition and medicinal chemistry, 24(3), 876-882 (2008-10-28)
Several substituted phenoxy acetic acid derived pyrazolines were synthesized by the reaction between 2-{4-[3-(2,4-dihydroxyphenyl)-3-oxo-1-propenyl]-2-methoxyphenoxy} acetic acid and substituted acid hydrazides and were tested for their in vitro cytotoxicity and antiviral activity. None of the compounds showed any specific antiviral activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service