Skip to Content
Merck
All Photos(2)

Key Documents

341088

Sigma-Aldrich

1,9-Dimethyl-Methylene Blue zinc chloride double salt

1,9-Dimethyl-Methylene Blue zinc chloride double salt

Dye content 80 %

Synonym(s):

DMMB, Taylor’s Blue

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22ClN3S · 0.5ZnCl2
CAS Number:
Molecular Weight:
416.05
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

form

powder

Quality Level

composition

Dye content, 80%

technique(s)

titration: suitable

impurities

<20% water

mp

250 °C (dec.) (lit.)

λmax

649 nm

ε (extinction coefficient)

≥10000 at 243-249 nm in H2O
≥18000 at 285-291 nm in H2O
≥25000 at 594-600 nm in H2O
≥35000 at 646-652 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cl-].Cl[Zn]Cl.CN(C)c1cc(C)c2N=C3C(C)=C\C(C=C3Sc2c1)=[N+](/C)C.CN(C)c4cc(C)c5N=C6C(C)=C\C(C=C6Sc5c4)=[N+](/C)C

InChI

1S/2C18H22N3S.4ClH.Zn/c2*1-11-7-13(20(3)4)9-15-17(11)19-18-12(2)8-14(21(5)6)10-16(18)22-15;;;;;/h2*7-10H,1-6H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

MKLBAKXCEQNMBC-UHFFFAOYSA-J

Application

1,9-Dimethyl-Methylene Blue zinc chloride double salt has been used for the quantification of sulfated glycosaminoglycans.[1]
DMMB is a cationic dye that specifically binds to sulfated glycosaminoglycans (sGAG) with an absorbance at ~525 nm.
DMMB is a cationic dye that specifically binds to sulfated glycosaminoglycans (sGAG) with an absorbance at ~525 nm. It has been used to quantitate glycosaminoglyans such as chondroitin sulfate, keratin sulfate, and heparan sulfate and sGAG chains in proteoglycans. One limitation to this method The sGAG-DMMB complex is unstable in solution and may precipitate. Precipitation of the complex was shown to improve stability.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi He et al.
FEBS open bio, 9(6), 1144-1152 (2019-05-01)
Osteoarthritis (OA) is a chronic degenerative disease that commonly affects the elderly. Current drug therapies for treating OA may cause adverse side effects, and so there remains a need to develop alternative treatments. Bergapten (BG) is a coumarin phytohormone that
P Behrendt et al.
Osteoarthritis and cartilage, 24(11), 1981-1988 (2016-10-21)
The aim of this study was to examine whether anti-inflammatory interleukin-10 (IL-10) exerts chondroprotective effects in an in vitro model of a single mechanical injury of mature articular cartilage. Articular cartilage was harvested from the femoro-patellar groove of adult cows (Bos
Anita Krouwels et al.
Spine, 43(5), 307-315 (2015-04-10)
An in vitro study using human degenerated nucleus pulposus cells. To determine the effect of osmolality and different osmolytes on the regeneration by human nucleus pulposus cells through gene expression and extracellular matrix production. Intervertebral disc (IVD) degeneration is a
Tianyi Wang et al.
Tissue engineering. Part A, 22(23-24), 1348-1356 (2016-10-21)
Cell-based therapies offer great promise for repairing cartilage. Previous strategies often involved using a single cell population such as stem cells or chondrocytes. A mixed cell population may offer an alternative strategy for cartilage regeneration while overcoming donor scarcity. We
Hyeon-Son Choi et al.
Biological & pharmaceutical bulletin, 39(12), 2042-2051 (2016-12-03)
The anti-osteoarthritic activity of the methanol fraction of deer bone oil extract (DBO-M) was evaluated in interleukin (IL)-1β-inflamed primary rabbit chondrocytes and in rats with monosodium iodoacetate (MIA)-induced osteoarthritis. The active compound in DBO-M was analyzed using a direct infusion

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service