Skip to Content
Merck
All Photos(1)

Key Documents

34042

Supelco

Rafoxanide

PESTANAL®, analytical standard

Synonym(s):

3′-Chloro-4′-(4-chlorophenoxy)-3,5-diiodosalicylanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H11Cl2I2NO3
CAS Number:
Molecular Weight:
626.01
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1c(I)cc(I)cc1C(=O)Nc2ccc(Oc3ccc(Cl)cc3)c(Cl)c2

InChI

1S/C19H11Cl2I2NO3/c20-10-1-4-13(5-2-10)27-17-6-3-12(9-15(17)21)24-19(26)14-7-11(22)8-16(23)18(14)25/h1-9,25H,(H,24,26)

InChI key

NEMNPWINWMHUMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rafoxanide is a halogenated salicylanilide and an anthelmintic agent. It is widely used against fasciolosis and haemonchosis in sheep and cattle. It also finds applications as an adulticide for Fasciola hepatica and Fasciola gigantica.

Application

Rafoxanide may be used as a reference standard in the determination of rafoxanide in plasma and tissue samples using high-performance liquid chromatography (HPLC) coupled with an ultraviolet detector and thermospray − mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

J H Gill et al.
International journal for parasitology, 25(4), 463-470 (1995-04-01)
Avermectin (AVM) inhibition of the development of the free-living stages of Haemonchus contortus has been quantified in an assay in which nematode eggs are placed on an agar matrix containing serial dilutions of a drug in the wells of a
J A van Wyk et al.
Veterinary parasitology, 72(2), 157-165 (1997-12-24)
We became increasingly concerned about indications of possible substandard efficacy of some generic anthelmintics, particularly after P.C. van Schalkwyk (personal communication, 1990) had found some batches of imported generic products obtained from international brokers to be poorly active, despite apparently
J A Van Wyk et al.
The Onderstepoort journal of veterinary research, 66(4), 273-284 (2000-02-26)
Surveys to determine the prevalence and degree of resistance of Haemonchus spp. of sheep and goats to the available anthelmintics in South Africa indicate that small ruminant production is entering a crisis situation. Three surveys employing the faecal egg count
R M Waruiru et al.
Veterinary parasitology, 75(2-3), 191-197 (1998-06-24)
The anthelmintic efficacy of benzimidazoles, levamisole, rafoxanide and ivermectin was evaluated on an institutional farm in Kenya using faecal egg count reduction test, larval cultures and a controlled slaughter trial. The results of this study indicated simultaneous resistance of Haemonchus
Veterinary Pharmacology and Therapeutics (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service