Skip to Content
Merck
All Photos(2)

Key Documents

32972

Supelco

Lactofen

PESTANAL®, analytical standard

Synonym(s):

2-Ethoxy-1-methyl-2-oxoethyl 5-(2-chloro-4-trifluoromethylphenoxy)-2-nitrobenzoate, Ethyl O-[5-(2-chloro-α,α,α-trifluoro-p-tolyloxy)-2-nitrobenzoyl]-DL-lactate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15ClF3NO7
CAS Number:
Molecular Weight:
461.77
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)C(C)OC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+]([O-])=O

InChI

1S/C19H15ClF3NO7/c1-3-29-17(25)10(2)30-18(26)13-9-12(5-6-15(13)24(27)28)31-16-7-4-11(8-14(16)20)19(21,22)23/h4-10H,3H2,1-2H3

InChI key

CONWAEURSVPLRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lactofen is a broad-spectrum post-emergent herbicide. It acts by inhibiting protoporphyrinogen oxidase by causing lipid peroxidation of the cell membrane.

Application

Lactofen may be used as an analytical reference standard for the determination of the analyte in:
  • Environmental samples by high performance liquid chromatography (HPLC) coupled to tandem mass spectrometric detector (MS/MS), ultraviolet (UV) detector and circular dichroism (CD) detector.
  • Fruit and wheat flour matrices by on-line turbulent flow chromatography (TFC) combined with LC-MS/MS.
  • Agricultural products using gas chromatography coupled to flame photometric detector (GC-FPD).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Madge Y Graham
Plant physiology, 139(4), 1784-1794 (2005-11-22)
Lactofen belongs to the diphenylether class of herbicides, which targets protoporphyrinogen oxidase, which in turn causes singlet oxygen generation. In tolerant plants like soybean (Glycine max), the chemical nonetheless causes necrotic patches called "bronzing" in contact areas. Here it is
Coupled turbulent flow chromatography: LC-MS/MS method for the analysis of pesticide residues in grapes, baby food and wheat flour matrices.
Hollosi L, et al.
Chromatographia, 75(23-24), 1377-1393 (2012)
Serena Landini et al.
Phytochemistry, 62(6), 865-874 (2003-02-19)
Lactofen, the active ingredient of the soybean disease resistance-inducing herbicide, Cobra, induces large accumulations of isoflavone conjugates and aglycones in soybean tissues. The predominant isoflavones induced in cotyledon tissues are daidzein (and its conjugates) and formononetin and glycitein aglycones. The
Irani Mukherjee et al.
Bulletin of environmental contamination and toxicology, 79(1), 49-52 (2007-06-30)
Lactofen, a member of the diphenyl ether chemical family, shows great potential for the control of broadleaf weeds associated with leguminous crops. It presents a high degree of selectivity when applied post-emergence to soybean and peanut crops. This paper presents
Bo Liang et al.
Journal of agricultural and food chemistry, 58(17), 9711-9715 (2010-08-18)
The diphenyl ether herbicide lactofen is commonly used to control broadleaf weeds. Once released into the environment, this herbicide is subject to microbial reactions. This study describes the biotransformation of lactofen by Brevundimonas sp. LY-2 isolated from enrichment cultures inoculated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service