Skip to Content
Merck
All Photos(1)

Key Documents

32541

Supelco

[D-Asp3, E-Dhb7]-Microcystin-RR solution

10 μg/mL in methanol, analytical standard

Synonym(s):

MCYST-AspDhbRR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H73N13O12
CAS Number:
Molecular Weight:
1024.17
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@@H](Cc1ccccc1)[C@@H](C)\C=C(C)\C=C\[C@@H]2NC(=O)[C@H](CCCNC(N)=N)NC(=O)C[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](C)NC(=O)\C(NC(=O)CC[C@@H](NC(=O)[C@H]2C)C(O)=O)=C/C)C(O)=O

InChI

1S/C48H73N13O12/c1-7-31-42(66)55-29(5)41(65)59-34(16-12-22-54-48(51)52)44(68)61-36(46(71)72)25-39(63)57-33(15-11-21-53-47(49)50)43(67)58-32(28(4)40(64)60-35(45(69)70)19-20-38(62)56-31)18-17-26(2)23-27(3)37(73-6)24-30-13-9-8-10-14-30/h7-10,13-14,17-18,23,27-29,32-37H,11-12,15-16,19-22,24-25H2,1-6H3,(H,55,66)(H,56,62)(H,57,63)(H,58,67)(H,59,65)(H,60,64)(H,61,68)(H,69,70)(H,71,72)(H4,49,50,53)(H4,51,52,54)/b18-17+,26-23+,31-7+/t27-,28-,29+,32-,33-,34-,35+,36+,37-/m0/s1

InChI key

JIVQGWHGELHXHV-UTGMKIGISA-N

General description

Microcystin is a type of toxin which can adversely affect human health. It is released by cyanobacteria and is commonly found in fresh water sources.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
[D-Asp3, E-Dhb7]-Microcystin-RR may be used as an analytical standard for the determination of the analyte in:
  • Raw water and drinking water samples by ultra-high pressure liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS).
  • Water samples, Planktothrix cultures and European crayfish by LC-MS.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC-MS analysis with thiol derivatization to differentiate [Dhb7]-from [Mdha7]-microcystins: analysis of cyanobacterial blooms, Planktothrix cultures and European crayfish from Lake Steinsfjorden, Norway.
Miles CO, et al.
Environmental Science & Technology, 47(9), 4080-4087 (2013)
Fast, rugged and sensitive ultra high pressure liquid chromatography tandem mass spectrometry method for analysis of cyanotoxins in raw water and drinking water-First findings of anatoxins, cylindrospermopsins and microcystin variants in Swedish source waters and infiltration ponds.
Pekar H, et al.
Journal of Chromatography A, 1429, 265-276 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service