Skip to Content
Merck
All Photos(2)

Key Documents

32406

Supelco

(±)-3-Chloro-1,2-propanediol

analytical standard

Synonym(s):

α-Chlorohydrin, α-Glycerol chlorohydrin, α-Monochlorohydrin, 3-MCPD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
110.54
Beilstein:
635684
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.04 mmHg ( 25 °C)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.480 (lit.)

bp

213 °C (lit.)

density

1.322 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OCC(O)CCl

InChI

1S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6H,1-2H2

InChI key

SSZWWUDQMAHNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(±)-3-Chloro-1,2-propanediol is a potent human carcinogen. It is formed as a product of acid hydrolysis of vegetable proteins during the process of cooking.

Application

(±)-3-Chloro-1,2-propanediol may be used as a reference standard for the determination of (±)-3-chloro-1,2-propanediol in food samples by gas chromatography with mass spectrometric detection (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 1

Target Organs

Kidney, Testes

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of 3-chloro-1, 2-propanediol in foods and food ingredients by gas chromatography with mass spectrometric detection: collaborative study.
Brereton P, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 84(2), 455-465 (2001)
Determination of 3-chloro-1, 2-propanediol in soy sauces by capillary electrophoresis with electrochemical detection.
Xing X and Cao Y
Food Control, 18(2), 167-172 (2007)
Youngbin Jang et al.
Food science and biotechnology, 29(12), 1665-1673 (2020-12-08)
Soy sauces collected from the Korean market were determined for 3-monochloropropane-1,2-diol (3-MCPD) and daily intake of 3-MCPD through the consumption of soy sauce was estimated. Thirty-one samples were chosen based on a market share in 2017. The amount of 3-MCPD varied
Brian D Craft et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(3), 354-361 (2011-12-16)
Recently, organic and inorganic chlorinated compounds were detected in crude and commercially refined palm oils. Further, the predominant formation mechanism of monochloropropanediol (MCPD) diesters at high temperatures (>170-180°C) was revealed. The present study involved the development and comparison of solutions
Z Zelinková et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 25(6), 669-676 (2008-05-20)
A series of twelve breast milk samples were analysed by gas chromatography-mass spectrometry (GC/MS) operated in selected ion monitoring mode for 3-chloropropane-1,2-diol (3-MCPD). Whilst none of the samples contained 3-MCPD above the limit of detection of 3 microg kg(-1) milk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service