Skip to Content
Merck
All Photos(2)

Key Documents

32403

Supelco

Cyflufenamid

PESTANAL®, analytical standard

Synonym(s):

(Z)-N-[α-(Cyclopropylmethoxyimino)-2,3-difluoro-6-(trifluoromethyl)benzyl]-2-phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17F5N2O2
CAS Number:
Molecular Weight:
412.35
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1ccc(c(c1F)C(\NC(=O)Cc2ccccc2)=N\OCC3CC3)C(F)(F)F

InChI

1S/C20H17F5N2O2/c21-15-9-8-14(20(23,24)25)17(18(15)22)19(27-29-11-13-6-7-13)26-16(28)10-12-4-2-1-3-5-12/h1-5,8-9,13H,6-7,10-11H2,(H,26,27,28)

InChI key

ACMXQHFNODYQAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cyflufenamid is a benzamidoxime fungicide, primarily used for the control of powdery mildew.

Application

Cyflufenamid may be used as an analytical reference standard for the determination of the analyte in honeybees and fruits/vegetables by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Thomas et al.
Plant methods, 14, 45-45 (2018-06-23)
Phenotyping is a bottleneck for the development of new plant cultivars. This study introduces a new hyperspectral phenotyping system, which combines the high throughput of canopy scale measurements with the advantages of high spatial resolution and a controlled measurement environment.
Recently Introduced Powdery Mildew Fungicides.
Dietz J and Winter C
Modern Crop Protection Compounds, 2, 933-947 (2019)
David Wari et al.
Pest management science, 76(9), 3236-3252 (2020-05-02)
Bemisia tabaci is a notorious agricultural pest that causes serious damage to many crops via herbivory and spread of viral diseases. Effective control measures are, therefore, required. Integrating entomopathogenic fungi into the chemical control system offers promise for B. tabaci
Timm Bernhard et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 132(5), 1335-1350 (2019-01-20)
The novel Rfm3 locus causing undesired fertility restoration in the msm1 cytoplasm of winter barley is located on the short arm of chromosome 6H. Undesired fertility restoration of cytoplasmic male sterile (CMS) mother lines in absence of the functional Rfm1
Vasileios Vangalis et al.
Journal of fungi (Basel, Switzerland), 6(4) (2020-12-11)
Woronin bodies are membrane-bound organelles of filamentous ascomycetes that mediate hyphal compartmentalization by plugging septal pores upon hyphal damage. Their major component is the peroxisomal protein Hex1, which has also been implicated in additional cellular processes in fungi. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service