Skip to Content
Merck
All Photos(1)

Key Documents

32344

Supelco

Cefpodoxime

VETRANAL®, analytical standard

Synonym(s):

(6R,7R)-7-{2-(2-Amino-4-thiazolyl)-2-[(Z)-methoxyimino]acetamido}-3-(methoxymethyl)-3-cephem-4-carboxylate, Cefpodoxime free acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17N5O6S2
CAS Number:
Molecular Weight:
427.46
Beilstein:
6021381
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

−20°C

SMILES string

O=C(/C(C1=CSC(N)=N1)=N\OC)N[C@H](C2=O)[C@]3([H])N2C(C(O)=O)=C(COC)CS3

InChI

1S/C15H17N5O6S2/c1-25-3-6-4-27-13-9(12(22)20(13)10(6)14(23)24)18-11(21)8(19-26-2)7-5-28-15(16)17-7/h5,9,13H,3-4H2,1-2H3,(H2,16,17)(H,18,21)(H,23,24)/b19-8-/t9-,13-/m1/s1

InChI key

WYUSVOMTXWRGEK-HBWVYFAYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axel Kola et al.
Antimicrobial resistance and infection control, 4, 8-8 (2015-03-31)
Carbapenemase-producing Enterobacteriaceae (CPE) have become a major problem for healthcare systems worldwide. While the first reports from European hospitals described the introduction of CPE from endemic countries, there is now a growing number of reports describing outbreaks of CPE in
Yen-Hung Liu et al.
International journal of antimicrobial agents, 51(5), 768-774 (2018-02-27)
A high prevalence of gonococcal resistance to various antimicrobials and Neisseria gonorrhoeae isolates exhibiting resistance to extended-spectrum cephalosporins have been reported in the past few decades. A total of 226 N. gonorrhoeae isolates obtained from the National Taiwan University Hospital
Vita Skuja et al.
European journal of gastroenterology & hepatology, 30(1), 92-100 (2017-10-28)
Certain Enterobacteriaceae strains have been associated with the development of ulcerative colitis (UC). Extended-spectrum β-lactamase (ESBL)-producing Enterobacteriaceae are the most commonly found multi-drug-resistant (MDR) bacteria colonizing the gut in UC patients and might trigger a more severe disease activity in
Leon Peto et al.
PloS one, 14(11), e0222831-e0222831 (2019-11-09)
Metagenomic sequencing of fecal DNA can usefully characterise an individual's intestinal resistome but is limited by its inability to detect important pathogens that may be present at low abundance, such as carbapenemase or extended-spectrum beta-lactamase producing Enterobacteriaceae. Here we aimed
Salome N Seiffert et al.
Frontiers in microbiology, 8, 407-407 (2017-04-01)
The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service