Skip to Content
Merck
All Photos(2)

Key Documents

32142

Supelco

Pirbuterol acetate

VETRANAL®, analytical standard

Synonym(s):

(RS)-2-(tert-Butylamino)-1-(6-hydroxymethyl-5-hydroxy-2-pyridyl)ethanol acetate salt, 6-(2-tert-Butylamino-1-hydroxyethyl)-2-hydroxymethylpyridin-3-ol acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N2O3 · C2H4O2
CAS Number:
Molecular Weight:
300.35
Beilstein:
13318829
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

InChI

1S/C12H20N2O3.C2H4O2/c1-12(2,3)13-6-11(17)8-4-5-10(16)9(7-15)14-8;1-2(3)4/h4-5,11,13,15-17H,6-7H2,1-3H3;1H3,(H,3,4)

InChI key

QSXMZJGGEWYVCN-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salbutamol, terbutaline and pirbuterol allergy in an asthmatic patient.
P Bonniaud et al.
Allergy, 62(10), 1219-1220 (2007-09-12)
F J Derksen et al.
Equine veterinary journal, 24(2), 107-112 (1992-03-01)
The dose of aerosol pirbuterol that could be administered safely to ponies (weight approximately 200 kg) was determined by observation for sweating, trembling and excitement and measurement of heart and respiratory rates during cumulative administration of the drug. Sweating, trembling
Makito Yaegashi et al.
Respiratory medicine, 101(5), 995-1000 (2006-10-24)
To address the utility of the forced oscillation technique (FOT) in assessing bronchodilator responsiveness compared with forced expiratory volume in 1s (FEV(1)). This is a retrospective analysis of consecutive 126 patients with a clinical history of asthma without any other
M H Schecker et al.
The Journal of allergy and clinical immunology, 92(6), 783-789 (1993-12-01)
Despite widespread acceptance of metered-dose inhalers (MDIs) in the treatment of asthma, many patients fail to operate these devices correctly. Inability to properly coordinate activation with onset of inhalation is regarded as the major factor in suboptimal MDI therapy. We
Z Ming et al.
Circulation, 95(6), 1568-1576 (1997-03-18)
We considered that beta 2-adrenergic stimulation may dilate resistance coronary vessels by opening ATP-sensitive potassium (KATP) channels, thereby triggering NO formation. In conscious instrumented dogs after beta 1-adrenergic blockade, intracoronary (IC) injections of acetylcholine (ACh), nitroglycerin (NTG), and pirbuterol (PIR)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service