Skip to Content
Merck
All Photos(1)

Key Documents

320579

Sigma-Aldrich

Xylenes

ACS reagent, ≥98.5% xylenes + ethylbenzene basis

Synonym(s):

Xylene mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

ACS reagent

vapor density

3.7 (vs air)

vapor pressure

18 mmHg ( 37.7 °C)

Assay

≥98.5% xylenes + ethylbenzene basis

form

liquid

autoignition temp.

867 °F

composition

C6H5CH2CH3, ≤25%

expl. lim.

7 %

impurities

H2SO4, passes test (substances darkened by sulfuric acid)
≤0.003% S compounds
≤0.05% water

evapn. residue

≤0.002%

color

APHA: ≤10

refractive index

n20/D 1.497 (lit.)

bp

137-140 °C (lit.)

mp

-34 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(C)cc1.Cc2cccc(C)c2.Cc3ccccc3C

InChI

1S/3C8H10/c1-7-3-5-8(2)6-4-7;1-7-4-3-5-8(2)6-7;1-7-5-3-4-6-8(7)2/h3*3-6H,1-2H3

InChI key

MVZVDAGWAAZJPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, hearing organs, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer V Littlejohns et al.
Journal of hazardous materials, 175(1-3), 872-882 (2009-12-05)
A dynamic mathematical model has been developed to predict the performance of a stirred tank, solid-liquid two-phase partitioning bioreactor (SL-TPPB) for the treatment of benzene, toluene, ethylbenzene and o-xylene (BTEX) contaminated gases. The SL-TPPB system consists of an aqueous phase
Cheng-Xiong Yang et al.
Analytical chemistry, 83(18), 7144-7150 (2011-08-04)
The diverse structures and pore topologies, accessible cages and tunnels, and high surface areas make metal-organic frameworks attractive as novel media in separation sciences. Here we report the slurry-packed MIL-101(Cr) column for high-performance liquid chromatographic separation of substituted aromatics. The
Zifu Li et al.
Chemical communications (Cambridge, England), 47(1), 331-333 (2010-08-24)
We describe a flexible method for large-scale production of polymeric microspheres by templating a microgel-stabilized oil-in-water (o/w) high internal phase emulsion.
Sailendra Nath Sarma et al.
Toxicology, 271(3), 122-130 (2010-04-03)
Benzene, toluene, o-xylene, ethylbenzene, trichloroethylene and dichloromethane are the most widely used volatile organic compounds (VOCs), and their toxic mechanisms are still undefined. This study analyzed the genome-wide expression profiles of human promyelocytic leukemia HL-60 cells exposed to VOCs using
Qiming Wang et al.
Nature communications, 5, 4899-4899 (2014-09-17)
Cephalopods can display dazzling patterns of colours by selectively contracting muscles to reversibly activate chromatophores--pigment-containing cells under their skins. Inspired by this novel colouring strategy found in nature, we design an electro-mechano-chemically responsive elastomer system that can exhibit a wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service