Skip to Content
Merck
All Photos(2)

Key Documents

31599

Supelco

Tebupirimfos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23N2O3PS
CAS Number:
Molecular Weight:
318.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=S)(OC(C)C)Oc1cnc(nc1)C(C)(C)C

InChI

1S/C13H23N2O3PS/c1-7-16-19(20,17-10(2)3)18-11-8-14-12(15-9-11)13(4,5)6/h8-10H,7H2,1-6H3

InChI key

AWYOMXWDGWUJHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tebupirimfos belongs to the class of organophosphorus pesticides (OPPs), which acts by inhibiting acetyl-cholinesterase enzyme activity. OPPs may be mutagens, carcinogens, and endocrine disruptors and can be harmful to human health and cause environmental contamination.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tebupirimfos may be used as a pesticide reference standard for the determination of the analyte:
  • In Sanjie Zhentong capsules, the traditional Chinese medicine based on a quick, easy, cheap, effective, rugged and safe (QuEChERS) method using ultra high performance liquid chromatography with tandem mass spectrometry.
  • In fruit and vegetable-based infant food by liquid chromatography/electrospray ionization coupled with tandem mass spectrometry.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of 35 ultra-trace level organophosphorus pesticide residues in Sanjie Zhentong capsules of traditional Chinese medicine using ultra high performance liquid chromatography with tandem mass spectrometry.
Li J, et al.
Journal of Separation Science, 40(4), 999- 1009 (2017)
Determination of 142 pesticides in fruit-and vegetable-based infant foods by liquid chromatography/electrospray ionization-tandem mass spectrometry and estimation of measurement uncertainty.
Wang J and Leung D
Journal of AOAC (Association of Official Analytical Chemists) International, 92(1), 279-301 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service