Skip to Content
Merck
All Photos(1)

Key Documents

30585

Supelco

Bis(1-butylpentyl)decane-1,10-diyl diglutarate

Selectophore

Synonym(s):

5,30-Dibutyl-6,12,23,29-tetraoxatetratriacontane-7,11,24,28-tetrane, ETH 469

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H70O8
CAS Number:
Molecular Weight:
654.96
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥97%

form

liquid

SMILES string

CCCCC(CCCC)OC(=O)CCCC(=O)OCCCCCCCCCCOC(=O)CCCC(=O)OC(CCCC)CCCC

InChI

1S/C38H70O8/c1-5-9-23-33(24-10-6-2)45-37(41)29-21-27-35(39)43-31-19-17-15-13-14-16-18-20-32-44-36(40)28-22-30-38(42)46-34(25-11-7-3)26-12-8-4/h33-34H,5-32H2,1-4H3

InChI key

AAEJKGTZDCFBEA-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Bis(1-butylpentyl)decane-1,10-diyl diglutarate, was used for the preparation of optode membranes, which are sodium selective and are based on neutral ionophore, primarily used for the characterization of total sodium concentration in human blood plasma. It was also used for the preparation of nitrite-selective membrane electrode, used for the potentiometric determination of NAD(P)H coenzymes.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Plasticizer of high lipophilicity

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Seiler et al.
Clinical chemistry, 37(8), 1350-1355 (1991-08-01)
Plasticized poly(vinyl chloride) membranes that incorporate a highly lipophilic sodium-selective neutral ionophore (ETH 4120) and novel H(+)-selective chromo-ionophores, lipophilic isologs of Nile Blue, are used in the competitive binding of Na+ and H+ for a reversible, optical determination of sodium
Potentiometric Determination of NAD (P) H Coenzymes Using a Nitrite-Selective Membrane Electrode and a Nitrate Reductase Enzyme and the Application to Glucose Assay in Human Serum.
Katsu T
Analytical Sciences, 15 (2), 135-139 (1999)
Bicarbonate-sensitive electrode based on planar thin membrane technology.
U Oesch et al.
Analytical chemistry, 59(17), 2131-2135 (1987-09-01)
Influence of the organic matrix on the properties of membrane coated ion sensor field-effect transistors
Kolytcheva, N.V., et al.
Sensors and Actuators B, Chemical, 58, 456-463 (1999)
Oesch, U.
Ion measurements in physiology and medicine, 42-42 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service